Merck
  • An Antibody Targeting ICOS Increases Intratumoral Cytotoxic to Regulatory T-cell Ratio and Induces Tumor Regression.
An Antibody Targeting ICOS Increases Intratumoral Cytotoxic to Regulatory T-cell Ratio and Induces Tumor Regression.
Cancer immunology research (2020-10-02)
Richard C A Sainson, Anil K Thotakura, Miha Kosmac, Gwenoline Borhis, Nahida Parveen, Rachael Kimber, Joana Carvalho, Simon J Henderson, Kerstin L Pryke, Tracey Okell, Siobhan O'Leary, Stuart Ball, Cassie Van Krinks, Lauriane Gamand, Emma Taggart, Eleanor J Pring, Hanif Ali, Hannah Craig, Vivian W Y Wong, Qi Liang, Robert J Rowlands, Morgane Lecointre, Jamie Campbell, Ian Kirby, David Melvin, Volker Germaschewski, Elisabeth Oelmann, Sonia Quaratino, Matthew McCourt
ABSTRACT
The immunosuppressive tumor microenvironment constitutes a significant hurdle to immune checkpoint inhibitor responses. Both soluble factors and specialized immune cells, such as regulatory T cells (Treg), are key components of active intratumoral immunosuppression. Inducible costimulatory receptor (ICOS) can be highly expressed in the tumor microenvironment, especially on immunosuppressive Treg, suggesting that it represents a relevant target for preferential depletion of these cells. Here, we performed immune profiling of samples from tumor-bearing mice and patients with cancer to demonstrate differential expression of ICOS in immune T-cell subsets in different tissues. ICOS expression was higher on intratumoral Treg than on effector CD8 T cells. In addition, by immunizing an Icos knockout transgenic mouse line expressing antibodies with human variable domains, we selected a fully human IgG1 antibody called KY1044 that bound ICOS from different species. We showed that KY1044 induced sustained depletion of ICOShigh T cells but was also associated with increased secretion of proinflammatory cytokines from ICOSlow effector T cells (Teff). In syngeneic mouse tumor models, KY1044 depleted ICOShigh Treg and increased the intratumoral TEff:Treg ratio, resulting in increased secretion of IFNγ and TNFα by TEff cells. KY1044 demonstrated monotherapy antitumor efficacy and improved anti-PD-L1 efficacy. In summary, we demonstrated that using KY1044, one can exploit the differential expression of ICOS on T-cell subtypes to improve the intratumoral immune contexture and restore an antitumor immune response.
Product Description
Corning® 96 well Polypropylene Deep Well Plate, V-bottom clear, polypropylene, bag of 10 ×, non-sterile, lid: no
Sigma-Aldrich
Red Blood Cell Lysing Buffer Hybri-Max, liquid, sterile-filtered, suitable for hybridoma