- Comment on "Synovial fibroblast-neutrophil interactions promote pathogenic adaptive immunity in rheumatoid arthritis".
Comment on "Synovial fibroblast-neutrophil interactions promote pathogenic adaptive immunity in rheumatoid arthritis".
Science immunology (2018-04-01)
Jing Shi, Felipe Andrade
PMID29602803
ABSTRACT
Discrepancies on the role of citrullination in the induction of experimental arthritis by neutrophils extracellular traps.