Merck
  • EMC3 Is Essential for Retinal Organization and Neurogenesis During Mouse Retinal Development.
EMC3 Is Essential for Retinal Organization and Neurogenesis During Mouse Retinal Development.
Investigative ophthalmology & visual science (2021-02-20)
Xiaowen Cao, Jianhong An, Yuqing Cao, Juan Lv, Jiawei Wang, Yang Ding, Xinhua Lin, Xiangtian Zhou
ABSTRACT
We used a mouse model to explore the role of the endoplasmic reticulum membrane protein complex subunit 3 (EMC3) in mammalian retinal development. The transcription pattern of Emc3 in C57BL/6 mice was analyzed by in situ hybridization. To explore the effects of EMC3 absence on retinal development, the Cre-loxP system was used to generate retina-specific Emc3 in knockout mice (Emc3flox/flox, Six3-cre+; CKO). Morphological changes in the retina of E13.5, E17.5, P0.5, and P7 mice were observed via hematoxylin and eosin staining. Immunofluorescence staining was used to assess protein distribution and terminal deoxynucleotidyl transferase dUTP nick end labeling (TUNEL) staining to assess apoptosis changes. Proteins were identified and quantified by Western blotting and proteomic analysis. Electroretinogram (ERG), fundus color photography, and optical coherence tomography were performed on 5-week-old mice to evaluate retinal function and structure. The Emc3 mRNA was widely distributed in the whole retina during development. Loss of retinal EMC3 led to retinal rosette degeneration with mislocalization of cell junction molecules (β-catenin, N-cadherin, and zonula occludens-1) and polarity molecules (Par3 and PKCζ). Endoplasmic reticulum stress and TUNEL apoptosis signals were present in retinal rosette-forming cells. Although the absence of EMC3 promoted the production of photoreceptor cells, 5-week-old mice lost all visual function and had severe retinal morphological degeneration. EMC3 regulates retinal structure by maintaining the polarity of retinal progenitor cells and regulating retinal cell apoptosis.
Sigma-Aldrich
Anti-phospho-Histone H3 (Ser10) Antibody, Mitosis Marker, Upstate®, from rabbit
Roche
In Situ Cell Death Detection Kit, TMR red, sufficient for ≤50 tests
Sigma-Aldrich
Anti-Prospero homeobox protein 1/PROX1 Antibody, from rabbit, purified by affinity chromatography