Merck
  • A Regulation Loop between YAP and NR4A1 Balances Cell Proliferation and Apoptosis.
A Regulation Loop between YAP and NR4A1 Balances Cell Proliferation and Apoptosis.
Cell reports (2020-10-22)
Lingli He, Liang Yuan, Wentao Yu, Yang Sun, Dan Jiang, Xiaodong Wang, Xue Feng, Zuoyun Wang, Jinjin Xu, Ruizeng Yang, Wenxiang Zhang, Hua Feng, Hang-Zi Chen, Yi Arial Zeng, Lijian Hui, Qiao Wu, Yonglong Zhang, Lei Zhang
ABSTRACT
The Hippo signaling pathway maintains organ size and tissue homeostasis via orchestration of cell proliferation and apoptosis. How this pathway triggers cell apoptosis remains largely unexplored. Here, we identify NR4A1 as a target of the Hippo pathway that mediates the pro-apoptotic and anti-tumor effects of the Hippo pathway whereby YAP regulates the transcription, phosphorylation, and mitochondrial localization of NR4A1. NR4A1, in turn, functions as a feedback inhibitor of YAP to promote its degradation, thereby inhibiting the function of YAP during liver regeneration and tumorigenesis. Our studies elucidate a regulatory loop between NR4A1 and YAP to coordinate Hippo signaling activity during liver regeneration and tumorigenesis and highlight NR4A1 as a marker of Hippo signaling, as well as a therapeutic target for hepatocellular carcinoma.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)