Merck
  • Suppression of miR-155 attenuates neuropathic pain by inducing an M1 to M2 switch in microglia.
Suppression of miR-155 attenuates neuropathic pain by inducing an M1 to M2 switch in microglia.
Folia neuropathologica (2020-04-28)
Yanxiang Zhang, Qi Chen, Yu Nai, Chunni Cao
ABSTRACT
The polarization state of microglia affects the progress of neuropathic pain. MiR-155 modulates polarization of microglia, while its role in neuropathic pain has not been well studied. We separately used lipopolysaccharide (LPS) and interleukin 4 (IL-4) for constructing an M1/M2 polarization model in BV-2 cells. The levels of CD86, iNOS, CD206, Arg and miR-155 were measured by western blot or qRT-PCR, as needed. Subsequently, BV-2 cells were transfected with miR-155 mimics or inhibitor to explore the effects of miR-155 on polarization states. We also constructed a neuropathic pain model by applying spinal nerve ligation (SNL) in Wistar rats with miR-155 agomir or antagomir injection. The withdrawal threshold was measured by Von Frey fibre needle. The levels of interleukin 1b (IL-1b), tumour necrosis factor a (TNF-a) and the proportion of M1-polarized microglia in primary microglia from rats were measured by ELISA and flow cytometry. LPS induced M1 polarization in BV-2 cells with increasing of CD86, iNOS and miR-155, while IL-4 induced M2 polarization in BV-2 cells with increasing of CD206, Arg and decreasing of miR-155. MiR-155 mimics upregulated CD86 and downregulated CD206, whereas miR-155 inhibitor downregulated CD86 and upregulated CD206. MiR-155 antagomir increased the withdrawal threshold, decreased the production of IL-1b, TNF-a and the proportion of M1-polarized microglia in primary microglia. Results demonstrate that suppression of miR-155 attenuates neuropathic pain by inducing an M1 to M2 switch in microglia. Our findings provide a new perspective to understand the function of miR-155 in microglia.
Sigma-Aldrich
Anti-Rabbit IgG, Native−Peroxidase antibody, Mouse monoclonal, clone RabT-50, purified from hybridoma cell culture
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Millipore
Phosphatase Inhibitor Cocktail Set III, Lyophilized, The Phosphatase Inhibitor Cocktail Set III, Lyophilized controls the activity of Phosphatase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.