Merck
Synthesis and trypanocidal activity of ent-kaurane glycosides.
Bioorganic & medicinal chemistry (2006-10-24)
Ronan Batista, Jorge Luiz Humberto, Egler Chiari, Alaíde Braga de Oliveira
ABSTRACT
Novel ent-kaurane glucosides were synthezised by a Koenigs-Knorr reaction between C17 and C19 alcohols derived from kaurenoic acid and 2,3,4,6-tetra-O-acetyl-glucopyranosyl bromide, followed by the hydrolysis of the acetates. Main products were assayed in vitro and in vivo against blood trypomastigote forms of Trypanosoma cruzi, the aetiological agent of Chagas' disease (American trypanosomiasis). The results allowed to establish structure-activity relationships among these derivatives, as well as pointed out the C19-methylester-C17-O-glucoside as a potential trypanocidal agent, whose trypanocidal profile was shown to be comparable to those of gentian violet and benznidazole.
Sigma-Aldrich
Gentian Violet, meets USP testing specifications
Supelco
Crystal Violet, VETRANAL®, analytical standard
Sigma-Aldrich
Crystal Violet, indicator for the determination of the redox potential, S. No.: 785
Sigma-Aldrich
Crystal Violet, for microscopy (Bact., Bot., Hist., Vit.), indicator (pH 0.1-2.0)
Sigma-Aldrich
Gentian violet, for microscopy (Bact., Hist.)
Sigma-Aldrich
Crystal Violet, certified by the Biological Stain Commission
Sigma-Aldrich
Crystal Violet, ACS reagent, ≥90.0% anhydrous basis