Merck
  • QSAR study on permeability of hydrophobic compounds with artificial membranes.
QSAR study on permeability of hydrophobic compounds with artificial membranes.
Bioorganic & medicinal chemistry (2007-04-10)
Masaaki Fujikawa, Kazuya Nakao, Ryo Shimizu, Miki Akamatsu
ABSTRACT
We previously reported a classical quantitative structure-activity relationship (QSAR) equation for permeability coefficients (P(app-pampa)) by parallel artificial membrane permeation assay (PAMPA) of structurally diverse compounds with simple physicochemical parameters, hydrophobicity at a particular pH (logP(oct) and |pK(a)-pH|), hydrogen-accepting ability (SA(HA)), and hydrogen-donating ability (SA(HD)); however, desipramine, imipramine, and testosterone, which have high logP(oct) values, were excluded from the derived QSAR equation because their measured P(app-pampa) values were lower than calculated. In this study, for further investigation of PAMPA permeability of hydrophobic compounds, we experimentally measured the P(app-pampa) of more compounds with high hydrophobicity, including several pesticides, and compared the measured P(app-pampa) values with those calculated from the QSAR equation. As a result, compounds having a calculated logP(app-pampa)>-4.5 showed lower measured logP(app-pampa) than calculated because of the barrier of the unstirred water layer and the membrane retention of hydrophobic compounds. The bilinear QSAR model explained the PAMPA permeability of the whole dataset of compounds, whether hydrophilic or hydrophobic, with the same parameters as the equation in the previous study. In addition, PAMPA permeability coefficients correlated well with Caco-2 cell permeability coefficients. Since Caco-2 cell permeability is effective for the evaluation of human oral absorption of compounds, the proposed bilinear model for PAMPA permeability could be useful for not only effective screening for several drug candidates but also the risk assessment of chemicals and agrochemicals absorbed by humans.
Sigma-Aldrich
o-Cresol, ReagentPlus®, ≥99%
Supelco
Pyrene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Melting point standard 235-237°C, analytical standard
Sigma-Aldrich
Indole-3-propionic acid, ≥99.0% (T)
Sigma-Aldrich
Diphenylamine, puriss. p.a., redox indicator, ACS reagent, reag. Ph. Eur., ≥98% (GC)
Supelco
Ibuprofen
Supelco
Diphenylamine, PESTANAL®, analytical standard
Supelco
4-Chloroaniline, PESTANAL®, analytical standard
Sigma-Aldrich
Indole-3-propionic acid, ≥97.0% (T)
Supelco
Dimethyl phthalate, PESTANAL®, analytical standard
Sigma-Aldrich
Salicylic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Aniline, ReagentPlus®, 99%
Sigma-Aldrich
o-Cresol, ≥99%
Sigma-Aldrich
Salicylic acid, meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance)
Supelco
Prednisolone, VETRANAL®, analytical standard
Supelco
Trimethoprim, VETRANAL®, analytical standard
Sigma-Aldrich
Pyrene, puriss. p.a., for fluorescence, ≥99.0% (GC)
Sigma-Aldrich
Dimethyl phthalate, ≥99%
Sigma-Aldrich
Trimethoprim, ≥99.0% (HPLC)
Sigma-Aldrich
Indole, ≥99%, FG
Sigma-Aldrich
4-Chloroaniline, purified by sublimation, ≥99%
Sigma-Aldrich
Diphenylamine, ACS reagent, ≥99%
Sigma-Aldrich
Aniline, ACS reagent, ≥99.5%
Sigma-Aldrich
4-Chloroaniline, 98%
Sigma-Aldrich
2,4-Dichlorophenol, 99%
Sigma-Aldrich
Corticosterone, ≥98.5% (HPLC)
Supelco
Fenobucarb, PESTANAL®, analytical standard
Pyrene, BCR®, certified reference material
Supelco
Aniline, analytical standard
Supelco
Pyrene, analytical standard