Merck
  • Generation of Functional Brown Adipocytes from Human Pluripotent Stem Cells via Progression through a Paraxial Mesoderm State.
Generation of Functional Brown Adipocytes from Human Pluripotent Stem Cells via Progression through a Paraxial Mesoderm State.
Cell stem cell (2020-08-14)
Liang Zhang, John Avery, Amelia Yin, Amar M Singh, Timothy S Cliff, Hang Yin, Stephen Dalton
ABSTRACT
Brown adipocytes (BAs) are a potential cell source for the treatment of metabolic disease, including type 2 diabetes. In this report, human pluripotent stem cells (hPSCs) are subject to directed differentiation through a paraxial mesoderm progenitor state that generates BAs at high efficiency. Molecular analysis identifies potential regulatory networks for BA development, giving insight into development along this lineage. hPSC-derived BAs undergo elevated rates of glycolysis, uncoupled respiration, and lipolysis that are responsive to changes in cyclic AMP (cAMP)-dependent signaling, consistent with metabolic activity in BA tissue depots. Transplanted human BAs engraft into the inter-scapular region of recipient mice and exhibit thermogenic activity. Recipient animals have elevated metabolic activity, respiratory exchange ratios, and whole-body energy expenditure. Finally, transplanted BAs reduce circulating glucose levels in hyperglycemic animals. These data provide a roadmap for brown adipocyte development and indicate that BAs generated from hPSCs have potential as a tool for therapeutic development.
Sigma-Aldrich
Free Glycerol Reagent, used for quantitative enzymatic determination of glycerol
Sigma-Aldrich
Glycerol Standard Solution, 2.5 mg/ml equivalent triolein concentration
Sigma-Aldrich
3-(Trimethylsilyl)-1-propanesulfonic acid sodium salt, 97%
Sigma-Aldrich
Paraformaldehyde, powder, 95%
Sigma-Aldrich
Sodium palmitate, ≥98.5%
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium, Without glucose, L-glutamine, phenol red, sodium pyruvate and sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
DL-Dithiothreitol, for molecular biology, ≥98% (HPLC), ≥99% (titration)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
TWEEN® 20, BioXtra, viscous liquid
Sigma-Aldrich
L-Ascorbic acid 2-phosphate sesquimagnesium salt hydrate, ≥95%
Sigma-Aldrich
Rosiglitazone, ≥98% (HPLC)
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
3,3′,5-Triiodo-L-thyronine sodium salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
L-Carnitine hydrochloride, synthetic, ≥98%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
RIPA Buffer
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial