Merck
  • Poly (Dimethylsiloxane) Coating for Repellency of Polar and Non-Polar Liquids.
Poly (Dimethylsiloxane) Coating for Repellency of Polar and Non-Polar Liquids.
Polymers (2020-10-25)
Hila Monder, Leo Bielenki, Hanna Dodiuk, Anna Dotan, Samuel Kenig
ABSTRACT
The wettability of poly (dimethylsiloxane) (PDMS) coating on plasma-treated glass was studied at room temperature using polar and non-polar liquids. The wettability was investigated regarding the liquids' surface tensions (STs), dielectric constants (DCs) and solubility parameters (SPs). For polar liquids, the contact angle (CA) and contact angle hysteresis (CAH) are controlled by the DCs and non-polar liquids by the liquids' STs. Solubility parameter difference between the PDMS and the liquids demonstrated that non-polar liquids possessed lower CAH. An empirical model that integrates the interfacial properties of liquid/PDMS has been composed. Accordingly, the difference between the SPs of PDMS and the liquid is the decisive factor affecting CAH, followed by the differences in DCs and STs. Moreover, the interaction between the DCs and the SPs is of importance to minimize CAH. It has been concluded that CAH, and not CA, is the decisive attribute for liquid repellency of PDMS coating.
Sigma-Aldrich
Diiodomethane, ReagentPlus®, 99%, contains copper as stabilizer
Sigma-Aldrich
Dimethoxydimethylsilane, 95%