Merck
  • Na, K-ATPase α3 is a death target of Alzheimer patient amyloid-β assembly.
Na, K-ATPase α3 is a death target of Alzheimer patient amyloid-β assembly.
Proceedings of the National Academy of Sciences of the United States of America (2015-08-01)
Takayuki Ohnishi, Masako Yanazawa, Tomoya Sasahara, Yasuki Kitamura, Hidekazu Hiroaki, Yugo Fukazawa, Isao Kii, Takashi Nishiyama, Akiyoshi Kakita, Hiroyuki Takeda, Akihide Takeuchi, Yoshie Arai, Akane Ito, Hitomi Komura, Hajime Hirao, Kaori Satomura, Masafumi Inoue, Shin-ichi Muramatsu, Ko Matsui, Mari Tada, Michio Sato, Eri Saijo, Yoshiki Shigemitsu, Satoko Sakai, Yoshitaka Umetsu, Natsuko Goda, Naomi Takino, Hitoshi Takahashi, Masatoshi Hagiwara, Tatsuya Sawasaki, Genji Iwasaki, Yu Nakamura, Yo-ichi Nabeshima, David B Teplow, Minako Hoshi
ABSTRACT
Neurodegeneration correlates with Alzheimer's disease (AD) symptoms, but the molecular identities of pathogenic amyloid β-protein (Aβ) oligomers and their targets, leading to neurodegeneration, remain unclear. Amylospheroids (ASPD) are AD patient-derived 10- to 15-nm spherical Aβ oligomers that cause selective degeneration of mature neurons. Here, we show that the ASPD target is neuron-specific Na(+)/K(+)-ATPase α3 subunit (NAKα3). ASPD-binding to NAKα3 impaired NAKα3-specific activity, activated N-type voltage-gated calcium channels, and caused mitochondrial calcium dyshomeostasis, tau abnormalities, and neurodegeneration. NMR and molecular modeling studies suggested that spherical ASPD contain N-terminal-Aβ-derived "thorns" responsible for target binding, which are distinct from low molecular-weight oligomers and dodecamers. The fourth extracellular loop (Ex4) region of NAKα3 encompassing Asn(879) and Trp(880) is essential for ASPD-NAKα3 interaction, because tetrapeptides mimicking this Ex4 region bound to the ASPD surface and blocked ASPD neurotoxicity. Our findings open up new possibilities for knowledge-based design of peptidomimetics that inhibit neurodegeneration in AD by blocking aberrant ASPD-NAKα3 interaction.
Product Description
Supelco
Poly(ethyleneimine) solution, analytical standard, 50 % (w/v) in H2O
Sigma-Aldrich
Sodium selenite, γ-irradiated, lyophilized powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Anti-Tau Mouse mAb (TAU-5), liquid, clone TAU-5, Calbiochem®
Sigma-Aldrich
Anti-Microtubule-Associated Protein 2 (MAP2) Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Monoclonal Anti-MAP2 antibody produced in mouse, clone HM-2, ascites fluid
Sigma-Aldrich
Anti-Na+K+ ATPase α-2 Antibody, serum, Upstate®
Sigma-Aldrich
Anti-Tau Antibody, clone Tau 7, clone Tau 7, from mouse
Sigma-Aldrich
Anti-Actin Antibody, clone C4, clone C4, Chemicon®, from mouse
Sigma-Aldrich
Anti-phospho-GSK3 (Tyr279/Tyr216) Antibody, clone 5G-2F, clone 5G-2F, Upstate®, from mouse