Merck
  • CD40 ligand and autoantigen are involved in the pathogenesis of low-grade B-cell lymphomas of mucosa-associated lymphoid tissue.
CD40 ligand and autoantigen are involved in the pathogenesis of low-grade B-cell lymphomas of mucosa-associated lymphoid tissue.
Developmental immunology (1998-11-14)
A Greiner, C Knörr, Y Qin, A Schultz, A Marx, R A Kroczek, H K Müller-Hermelink
ABSTRACT
Low-grade MALT-type lymphomas are malignancies of mucosal marginal-zone B cells and preceded by reactive inflammatory lymphoid tissue. Experimental observations suggest that antigen and CD40 Ligand act during cognate T/B cell interaction and are crucial for germinal center B-cell maturation generating marginal-zone B cells. To investigate the mechanisms underlying the development of extranodal MALT-type lymphomas, the immunoglobulin receptor was sequenced and analyzed for antigen specificity using heterohybridoma technology. Furthermore, CD40 ligand expression was evaluated by immunohistochemistry and by semiquantitative RT-PCR, and ligand binding to the CD40 of tumor B cells was studied using the CD40 system. Hypermutations were found in low-grade lymphomas throughout CDR1-CDR3 suggestive of positive selection through their antigen receptor. Different VH families were used and more than 69% of tumor immunoglobulins bound different mucosal antigens. CD40L expression was found in the tumor marginal zone in substantial amounts. The in vitro proliferation response of all low-grade MALT-type lymphomas was dependent on anti-CD40-mediated signals and cytokines. Our data provide evidence that autoantigen as well as the CD40L expressed by activated nonneoplastic T cells may drive the evolution of low-grade MALT-type lymphomas either directly or by paracrine mechanisms and that antigen may contribute to lymphoma pathogenesis.
Sigma-Aldrich
Anti-CD19 Antibody, clone HD37, clone HD37, Chemicon®, from mouse