Merck
  • Myocardial infarction affects Cx43 content of extracellular vesicles secreted by cardiomyocytes.
Myocardial infarction affects Cx43 content of extracellular vesicles secreted by cardiomyocytes.
Life science alliance (2020-10-25)
Tania Martins-Marques, Teresa Ribeiro-Rodrigues, Saskia C de Jager, Monica Zuzarte, Cátia Ferreira, Pedro Cruz, Liliana Reis, Rui Baptista, Lino Gonçalves, Joost Pg Sluijter, Henrique Girao
ABSTRACT
Ischemic heart disease has been associated with an impairment on intercellular communication mediated by both gap junctions and extracellular vesicles. We have previously shown that connexin 43 (Cx43), the main ventricular gap junction protein, assembles into channels at the extracellular vesicle surface, mediating the release of vesicle content into target cells. Here, using a comprehensive strategy that included cell-based approaches, animal models and human patients, we demonstrate that myocardial ischemia impairs the secretion of Cx43 into circulating, intracardiac and cardiomyocyte-derived vesicles. In addition, we show that ubiquitin signals Cx43 release in basal conditions but appears to be dispensable during ischemia, suggesting an interplay between ischemia-induced Cx43 degradation and secretion. Overall, this study constitutes a step forward for the characterization of the signals and molecular players underlying vesicle protein sorting, with strong implications on long-range intercellular communication, paving the way towards the development of innovative diagnostic and therapeutic strategies for cardiovascular disorders.
Sigma-Aldrich
MISSION® esiRNA, targeting human USP8
Sigma-Aldrich
Anti-Hsp90 Mouse mAb (AC88), liquid, clone AC88, Calbiochem®