Merck
  • Simple criterion for selection of flavonoid compounds with anti-HIV activity.
Simple criterion for selection of flavonoid compounds with anti-HIV activity.
Bioorganic & medicinal chemistry letters (2006-12-27)
Veljko Veljkovic, Jean-François Mouscadet, Nevena Veljkovic, Sanja Glisic, Zeger Debyser
ABSTRACT
Flavonoid compounds represent an important natural source of antiretrovirals for AIDS therapy due to their significant anti-HIV-1 activity and low toxicity. Here we propose a simple theoretical criterion to discriminate active from inactive flavonoids that is suitable for rapid in silico screening of flavonoid libraries, and selection and optimization of lead compounds with anti-HIV-1 activity.
Sigma-Aldrich
Apigenin, ≥95.0% (HPLC)
Sigma-Aldrich
Kaempferol, ≥90% (HPLC), powder
Sigma-Aldrich
Kaempferol, ≥97.0% (HPLC)
Sigma-Aldrich
Myricetin, ≥96.0%, crystalline
Sigma-Aldrich
Myricetin, ≥96.0% (HPLC)
Sigma-Aldrich
Chrysin, ≥96.5%