Merck
  • Coupling substrate and ion binding to extracellular gate of a sodium-dependent aspartate transporter.
Coupling substrate and ion binding to extracellular gate of a sodium-dependent aspartate transporter.
Nature (2007-01-19)
Olga Boudker, Renae M Ryan, Dinesh Yernool, Keiko Shimamoto, Eric Gouaux
ABSTRACT
Secondary transporters are integral membrane proteins that catalyse the movement of substrate molecules across the lipid bilayer by coupling substrate transport to one or more ion gradients, thereby providing a mechanism for the concentrative uptake of substrates. Here we describe crystallographic and thermodynamic studies of Glt(Ph), a sodium (Na+)-coupled aspartate transporter, defining sites for aspartate, two sodium ions and d,l-threo-beta-benzyloxyaspartate, an inhibitor. We further show that helical hairpin 2 is the extracellular gate that controls access of substrate and ions to the internal binding sites. At least two sodium ions bind in close proximity to the substrate and these sodium-binding sites, together with the sodium-binding sites in another sodium-coupled transporter, LeuT, define an unwound alpha-helix as the central element of the ion-binding motif, a motif well suited to the binding of sodium and to participation in conformational changes that accompany ion binding and unbinding during the transport cycle.
Sigma-Aldrich
L-Aspartic acid, BioUltra, ≥99.5% (T)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
D-Aspartic acid, ReagentPlus®, 99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Aspartic acid, reagent grade, ≥98% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Aspartic acid, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
SKU
Pack Size
Availability
Price
Quantity
SAFC
L-Aspartic acid
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Aspartic acid, BioXtra, ≥99% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Supelco
L-Aspartic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Glutamic acid, FCC
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Succinic acid, ACS reagent, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Glutamic acid, BioUltra, ≥99.5% (NT)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Succinic acid, purum p.a., ≥99.0% (T)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Serine, ReagentPlus®, ≥99% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Serine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Succinic acid, BioXtra, BioRenewable, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Glutamic acid, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-100.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Succinic acid, BioReagent, BioRenewable, suitable for cell culture, suitable for insect cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Glutamic acid, ReagentPlus®, ≥99% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Succinic acid, ReagentPlus®, BioRenewable, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
D-Glutamic acid, ≥99% (TLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Serine, BioUltra, ≥99.5% (NT)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Succinic acid, puriss. p.a., ACS reagent, ≥99.5% (T)
SKU
Pack Size
Availability
Price
Quantity
Supelco
L-Glutamic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
SKU
Pack Size
Availability
Price
Quantity
Supelco
L-Serine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland