Merck
Poorly soluble marketed drugs display solvation limited solubility.
Journal of medicinal chemistry (2007-10-13)
Christel A S Bergström, Carola M Wassvik, Kajsa Johansson, Ina Hubatsch
ABSTRACT
We determined the intrinsic aqueous solubility of 15 poorly soluble drugs with solubilities ranging from 2.9 nM to 1.1 microM. We then analyzed the data from a physicochemical perspective, using experimentally determined solid-state properties and easily interpretable two-dimensional molecular descriptors, to better understand the factors underlying poor solubility. The analysis shows that poorly soluble drugs that have reached the market are solubility limited by solvation rather than by their solid state.
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Supelco
Albendazole, analytical standard, ≥98%
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Tolfenamic acid, NSAID
Sigma-Aldrich
Glyburide, meets USP testing specifications