Merck
  • Thioredoxin protects mitochondrial structure, function and biogenesis in myocardial ischemia-reperfusion via redox-dependent activation of AKT-CREB- PGC1α pathway in aged mice.
Thioredoxin protects mitochondrial structure, function and biogenesis in myocardial ischemia-reperfusion via redox-dependent activation of AKT-CREB- PGC1α pathway in aged mice.
Aging (2020-10-14)
Jaganathan Subramani, Venkatesh Kundumani-Sridharan, Kumuda C Das
ABSTRACT
Aging is an independent risk factor for cardiovascular diseases, such as myocardial infarction due to ischemia-reperfusion injury (I/R) of the heart. Cytosolic thioredoxin (Trx) is a multifunctional redox protein which has antioxidant and protein disulfide reducing properties. We hypothesized that high levels of Trx will protect against multifactorial disease such as myocardial infarction due to I/R injury in aged mice. Aged mice overexpressing human Trx (Trx-Tg), mice expressing redox-inactive mutant of human Trx (dnTrx-Tg) and non-transgenic litter-mates (NT) were subjected to I/R (60/30 min), and cardiac function, mitochondrial structure and function, and biogenesis involving PGC1α pathway were evaluated in these mice. While aged Trx-Tg mice were protected from I/R-induced reduction in ejection fraction (EF) and fractional shortening (FS), had smaller infarct with decreased apoptosis and preserved mitochondrial function, aged dnTrx-Tg mice showed enhanced myocardial injury and mitochondrial dysfunction. Further, Trx-Tg mice were protected from I/R induced loss of PGC1α, ACO2, MFN1 and MFN2 in the myocardium. The dnTrx-Tg mice were highly sensitive to I/R induced apoptosis. Overall, our study demonstrated that the loss of Trx redox balance in I/R in aged NT or dnTrx-Tg mice resulted in decreased PGC1α expression that decreased mitochondrial gene expression with increased myocardial apoptosis. High levels of Trx, but not mitochondrial thioredoxin (Trx-2) maintained Trx redox balance in I/R resulting in increased PGC1α expression via AKT/CREB activation upregulating mitochondrial gene expression and protection against I/R injury.
Product Description
Supelco
SUPELCOSIL LC-18-DB HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
Sigma-Aldrich
Anti-Mn-SOD Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-8-Oxoguanine Antibody, clone 483.15, ascites fluid, clone 483.15, Chemicon®