Merck
  • RIP1/RIP3/MLKL-mediated necroptosis contributes to vinblastine-induced myocardial damage.
RIP1/RIP3/MLKL-mediated necroptosis contributes to vinblastine-induced myocardial damage.
Molecular and cellular biochemistry (2020-11-29)
Huiling Zhou, Lijun Liu, Xiaolong Ma, Jian Wang, Jinfu Yang, Xinmin Zhou, Yifeng Yang, Haidan Liu
ABSTRACT
Vinblastine (VBL) has been considered as a first-line anti-tumor drug for many years. However, vinblastine-caused myocardial damage has been continually reported. The underlying molecular mechanism of the myocardial damage remains unknown. Here, we show that vinblastine induces myocardial damage and necroptosis is involved in the vinblastine-induced myocardial damage both in vitro and in vivo. The results of WST-8 and flow cytometry analysis show that vinblastine causes damage to H9c2 cells, and the results of animal experiments show that vinblastine causes myocardial cell damage. The necrosome components, receptor-interacting protein 1 (RIP1) receptor-interacting protein 3 (RIP3), are significantly increased in vinblastine-treated H9c2 cells, primary neonatal rat ventricular myocytes and rat heart tissues. And the downstream substrate of RIP3, mixed lineage kinase domain like protein (MLKL) was also increased. Pre-treatment with necroptosis inhibitors partially inhibits the necrosome components and MLKL levels and alleviates vinblastine-induced myocardial injury both in vitro and in vivo. This study indicates that necroptosis participated in vinblastine-evoked myocardial cell death partially, which would be a potential target for relieving the chemotherapy-related myocardial damage.
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Sigma-Aldrich
5-Bromo-2′-deoxyuridine, ≥99% (HPLC)
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
Sigma-Aldrich
2,3-Butanedione monoxime, ≥98%
Sigma-Aldrich
Normal Rabbit IgG, This Normal Rabbit IgG is validated for use as a negative control in parallel with specific primary antibodies in ELISA, FC, Immunoblotting, IF, IHC, IP.
Sigma-Aldrich
Anti-oxidized-CaM Kinase II (Met281/282) Antibody, serum, from rabbit