Merck
  • Endogenous Rab29 does not impact basal or stimulated LRRK2 pathway activity.
Endogenous Rab29 does not impact basal or stimulated LRRK2 pathway activity.
The Biochemical journal (2020-11-03)
Alexia F Kalogeropulou, Jordana B Freemantle, Pawel Lis, Edmundo G Vides, Nicole K Polinski, Dario R Alessi
ABSTRACT
Mutations that enhance LRRK2 protein kinase activity cause inherited Parkinson's disease. LRRK2 phosphorylates a group of Rab GTPase proteins, including Rab10 and Rab12, within the effector-binding switch-II motif. Previous work has indicated that the PARK16 locus, which harbors the gene encoding for Rab29, is involved in Parkinson's, and that Rab29 operates in a common pathway with LRRK2. Co-expression of Rab29 and LRRK2 stimulates LRRK2 activity by recruiting LRRK2 to the surface of the trans Golgi network. Here, we report that knock-out of Rab29 does not influence endogenous LRRK2 activity, based on the assessment of Rab10 and Rab12 phosphorylation, in wild-type LRRK2, LRRK2[R1441C] or VPS35[D620N] knock-in mouse tissues and primary cell lines, including brain extracts and embryonic fibroblasts. We find that in brain extracts, Rab12 phosphorylation is more robustly impacted by LRRK2 inhibitors and pathogenic mutations than Rab10 phosphorylation. Transgenic overexpression of Rab29 in a mouse model was also insufficient to stimulate basal LRRK2 activity. We observed that stimulation of Rab10 and Rab12 phosphorylation induced by agents that stress the endolysosomal system (nigericin, monensin, chloroquine and LLOMe) is suppressed by LRRK2 inhibitors but not blocked in Rab29 deficient cells. From the agents tested, nigericin induced the greatest increase in Rab10 and Rab12 phosphorylation (5 to 9-fold). Our findings indicate that basal, pathogenic, as well as nigericin and monensin stimulated LRRK2 pathway activity is not controlled by Rab29. Further work is required to establish how LRRK2 activity is regulated, and whether other Rab proteins can control LRRK2 by targeting it to diverse membranes.
Sigma-Aldrich
Leu-Leu methyl ester hydrobromide, ≥97% (TLC)
Sigma-Aldrich
Monensin sodium salt, 90-95% (TLC)
Sigma-Aldrich
Chloroquine diphosphate salt, powder or crystals, 98.5-101.0% (EP), meets EP testing specifications
Sigma-Aldrich
bisBenzimide H 33342 trihydrochloride, ≥98% (HPLC)
Sigma-Aldrich
(2-Hydroxypropyl)-β-cyclodextrin, average Mw ~1,460
Sigma-Aldrich
MISSION® esiRNA, targeting human LRRK2
Supelco
Microcystin-LR solution, 10 μg/mL in methanol, analytical standard