Merck
  • Extracellular clusterin limits the uptake of α-synuclein fibrils by murine and human astrocytes.
Extracellular clusterin limits the uptake of α-synuclein fibrils by murine and human astrocytes.
Glia (2020-10-13)
Alice Filippini, Veronica Mutti, Gaia Faustini, Francesca Longhena, Ileana Ramazzina, Federica Rizzi, Alice Kaganovich, Dorien A Roosen, Natalie Landeck, Megan Duffy, Isabella Tessari, Federica Bono, Chiara Fiorentini, Elisa Greggio, Luigi Bubacco, Arianna Bellucci, Mariacristina Missale, Mark R Cookson, Massimo Gennarelli, Isabella Russo
ABSTRACT
The progressive neuropathological damage seen in Parkinson's disease (PD) is thought to be related to the spreading of aggregated forms of α-synuclein. Clearance of extracellular α-synuclein released by degenerating neurons may be therefore a key mechanism to control the concentration of α-synuclein in the extracellular space. Several molecular chaperones control misfolded protein accumulation in the extracellular compartment. Among these, clusterin, a glycoprotein associated with Alzheimer's disease, binds α-synuclein aggregated species and is present in Lewy bodies, intraneuronal aggregates mainly composed by fibrillary α-synuclein. In this study, using murine primary astrocytes with clusterin genetic deletion, human-induced pluripotent stem cell (iPSC)-derived astrocytes with clusterin silencing and two animal models relevant for PD we explore how clusterin affects the clearance of α-synuclein aggregates by astrocytes. Our findings showed that astrocytes take up α-synuclein preformed fibrils (pffs) through dynamin-dependent endocytosis and that clusterin levels are modulated in the culture media of cells upon α-synuclein pffs exposure. Specifically, we found that clusterin interacts with α-synuclein pffs in the extracellular compartment and the clusterin/α-synuclein complex can be internalized by astrocytes. Mechanistically, using clusterin knock-out primary astrocytes and clusterin knock-down hiPSC-derived astrocytes we observed that clusterin limits the uptake of α-synuclein pffs by cells. Interestingly, we detected increased levels of clusterin in the adeno-associated virus- and the α-synuclein pffs- injected mouse model, suggesting a crucial role of this chaperone in the pathogenesis of PD. Overall, our observations indicate that clusterin can limit the uptake of extracellular α-synuclein aggregates by astrocytes and, hence, contribute to the spreading of Parkinson pathology.
Sigma-Aldrich
MISSION® esiRNA, targeting human CLU
Sigma-Aldrich
Triton X-100, laboratory grade