Merck
  • Acetylation-mediated remodeling of the nucleolus regulates cellular acetyl-CoA responses.
Acetylation-mediated remodeling of the nucleolus regulates cellular acetyl-CoA responses.
PLoS biology (2020-12-01)
Ryan Houston, Shiori Sekine, Michael J Calderon, Fayaz Seifuddin, Guanghui Wang, Hiroyuki Kawagishi, Daniela A Malide, Yuesheng Li, Marjan Gucek, Mehdi Pirooznia, Alissa J Nelson, Matthew P Stokes, Jacob Stewart-Ornstein, Steven J Mullett, Stacy G Wendell, Simon C Watkins, Toren Finkel, Yusuke Sekine
ABSTRACT
The metabolite acetyl-coenzyme A (acetyl-CoA) serves as an essential element for a wide range of cellular functions including adenosine triphosphate (ATP) production, lipid synthesis, and protein acetylation. Intracellular acetyl-CoA concentrations are associated with nutrient availability, but the mechanisms by which a cell responds to fluctuations in acetyl-CoA levels remain elusive. Here, we generate a cell system to selectively manipulate the nucleo-cytoplasmic levels of acetyl-CoA using clustered regularly interspaced short palindromic repeat (CRISPR)-mediated gene editing and acetate supplementation of the culture media. Using this system and quantitative omics analyses, we demonstrate that acetyl-CoA depletion alters the integrity of the nucleolus, impairing ribosomal RNA synthesis and evoking the ribosomal protein-dependent activation of p53. This nucleolar remodeling appears to be mediated through the class IIa histone deacetylases (HDACs). Our findings highlight acetylation-mediated control of the nucleolus as an important hub linking acetyl-CoA fluctuations to cellular stress responses.
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ~98% (HPLC)
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Nα-Acetyl-L-lysine
Sigma-Aldrich
Coenzyme A sodium salt hydrate, cofactor for acyl transfer
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Acetyl coenzyme A trisodium salt, ≥93% (HPLC), powder
Sigma-Aldrich
SAHA, ≥98% (HPLC)
Sigma-Aldrich
Trichostatin A, Ready Made Solution, 5 mM in DMSO, from Streptomyces sp.