Merck
  • Differential activation of the calpain system involved in individualized adaptation of different fast-twitch muscles in hibernating Daurian ground squirrels.
Differential activation of the calpain system involved in individualized adaptation of different fast-twitch muscles in hibernating Daurian ground squirrels.
Journal of applied physiology (Bethesda, Md. : 1985) (2019-06-21)
Xiufeng Ma, Hui Chang, Zhe Wang, Shenhui Xu, Xin Peng, Jie Zhang, Xia Yan, Tingyun Lei, Huiping Wang, Yunfang Gao
ABSTRACT
We examined the lateral gastrocnemius (LG), plantaris (PL), and extensor digitorum longus (EDL) muscles to determine whether differential activation of the calpain system is related to the degree of atrophy in these fast-twitch skeletal muscles during hibernation in Daurian ground squirrels (Spermophilus dauricus). Results from morphological indices showed various degrees of atrophy in the order LG > PL > EDL. Furthermore, all three muscles underwent fast-to-slow fiber-type conversion in hibernation. In regard to the calpain system in the LG muscle, cytosolic Ca2+ increased significantly in hibernation, followed by recovery in posthibernation. Furthermore, calpastatin expression significantly decreased, and calpain 1 and 2 expression significantly increased, which may be responsible for the increased degradation of desmin during hibernation compared with that during summer activity. In the EDL muscle, Ca2+ overload was observed during interbout arousal, and calpastatin showed an increase during hibernation and interbout arousal, which could explain the increased levels of troponin T during both periods compared with levels during summer activity. These findings suggest that cytosolic Ca2+ overload and subsequent calpain 1 and 2 activation may be an important mechanism of LG muscle atrophy during hibernation. Cytosolic Ca2+ homeostasis and high expression of calpain inhibitor calpastatin during hibernation may also be an important mechanism for the EDL muscle to maintain muscle mass. Thus, the differential activation of the calpain system and selective degradation of downstream substrates may be involved in muscle atrophy of different fast-twitch muscles during hibernation.NEW & NOTEWORTHY We found that the extent of both muscle atrophy and calpain system activation differed in fast-twitch lateral gastrocnemius (LG), plantaris (PL), and extensor digitorum longus (EDL) skeletal muscles in hibernating Daurian ground squirrels, but similar hierarchies in the order of LG > PL > EDL. The differential activation of the calpain system and selective degradation of downstream substrates may be involved in muscle atrophy in different fast-twitch muscles during hibernation.
Sigma-Aldrich
Anti-Mouse Polyvalent Immunoglobulins (G,A,M)−FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution