Merck
  • Specific Effects of Trabectedin and Lurbinectedin on Human Macrophage Function and Fate-Novel Insights.
Specific Effects of Trabectedin and Lurbinectedin on Human Macrophage Function and Fate-Novel Insights.
Cancers (2020-10-24)
Adrián Povo-Retana, Marina Mojena, Adrian B Stremtan, Victoria B Fernández-García, Ana Gómez-Sáez, Cristina Nuevo-Tapioles, José M Molina-Guijarro, José Avendaño-Ortiz, José M Cuezva, Eduardo López-Collazo, Juan F Martínez-Leal, Lisardo Boscá
ABSTRACT
Tumor-associated macrophages (TAMs) play a crucial role in suppressing the immunosurveillance function of the immune system that prevents tumor growth. Indeed, macrophages can also be targeted by different chemotherapeutic agents improving the action over immune checkpoints to fight cancer. Here we describe the effect of trabectedin and lurbinectedin on human macrophage cell viability and function. Blood monocytes from healthy donors were differentiated into macrophages and exposed to different stimuli promoting functional polarization and differentiation into tumor-associated macrophages. Cells were challenged with the chemotherapeutic drugs and the effects on cell viability and function were analyzed. Human macrophages exhibit at least two different profiles in response to these drugs. One-fourth of the blood donors assayed (164 individuals) were extremely sensitive to trabectedin and lurbinectedin, which promoted apoptotic cell death. Macrophages from other individuals retained viability but responded to the drugs increasing reactive oxygen production and showing a rapid intracellular calcium rise and a loss of mitochondrial oxygen consumption. Cell-membrane exposure of programmed-death ligand 1 (PD-L1) significantly decreased after treatment with therapeutic doses of these drugs, including changes in the gene expression profile of hypoxia-inducible factor 1 alpha (HIF-1α)-dependent genes, among other. The results provide evidence of additional onco-therapeutic actions for these drugs.
Sigma-Aldrich
TOFA, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl fumarate, 97%
Pricing and availability is not currently available.
Sigma-Aldrich
Rapamycin from Streptomyces hygroscopicus, ≥95% (HPLC), powder
Pricing and availability is not currently available.
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
Pricing and availability is not currently available.
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Pricing and availability is not currently available.
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Pricing and availability is not currently available.
SAFC
Insulin, Human Recombinant, for research or for further manufacturing use, dry powder
Pricing and availability is not currently available.
Sigma-Aldrich
CP-456773 sodium salt, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
3PO, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 3, DMSO solution
Pricing and availability is not currently available.
Sigma-Aldrich
Rosiglitazone, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium L-lactate, ~98%
Pricing and availability is not currently available.
Sigma-Aldrich
Lactacystin, ≥90% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium succinate dibasic hexahydrate, BioReagent, suitable for cell culture, suitable for insect cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Pricing and availability is not currently available.
Sigma-Aldrich
CHAPS hydrate, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
NF-κB SN50, Cell-Permeable Inhibitor Peptide, NF-κB SN50, CAS 213546-43-3, is a cell-permeable peptide that contains the NLS sequence of NF-κB p50 linked to the hydrophobic region of K-FGF. Inhibits translocation of the NF-κB into the nucleus.
Pricing and availability is not currently available.