Merck
  • Shank2 Binds to aPKC and Controls Tight Junction Formation with Rap1 Signaling during Establishment of Epithelial Cell Polarity.
Shank2 Binds to aPKC and Controls Tight Junction Formation with Rap1 Signaling during Establishment of Epithelial Cell Polarity.
Cell reports (2020-04-09)
Kazunori Sasaki, Noriko Kojitani, Hiroko Hirose, Yohei Yoshihama, Hidefumi Suzuki, Miho Shimada, Ayumi Takayanagi, Akio Yamashita, Masa-Aki Nakaya, Hisashi Hirano, Hidehisa Takahashi, Shigeo Ohno
ABSTRACT
Epithelial cells establish apicobasal polarity by forming tight junctions (TJs) at the apical-lateral boundary, which play fundamental roles in physiological functions. An evolutionarily conserved atypical protein kinase C (aPKC)-partitioning defective (PAR) complex functions as a platform for TJ assembly during cell polarity establishment. However, how this complex converts the spatial cues into a subsequent active unit is unclear. Here, we identify an epithelial isoform of Shank2 as a mediator of the aPKC-PAR complex. Shank2 binds to and colocalizes with aPKC at apical junctional regions of polarized epithelial cells. Shank2 knockdown results in defects in TJ formation. Mechanistically, we find that the N-terminal SPN domain is required for the junctional localization of Shank2 and binds to the active form of Rap1 small GTPase, which is involved in TJ formation. Our findings suggest that a close physical and functional relationship between aPKC and Shank2-active Rap1 signaling serves as the platform for TJ assembly to regulate epithelial cell polarity.
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Rap1 Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-Partitioning-defective 3 Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-E-cadherin Antibody, clone DECMA-1, clone Decma-1, from rat
Sigma-Aldrich
Anti-Giα3 Antibody, serum, Upstate®