Merck
  • 2H-benzimidazole 1,3-dioxide derivatives: a new family of water-soluble anti-trypanosomatid agents.
2H-benzimidazole 1,3-dioxide derivatives: a new family of water-soluble anti-trypanosomatid agents.
Journal of medicinal chemistry (2006-05-26)
Mariana Boiani, Lucía Boiani, Ana Denicola, Susana Torres de Ortiz, Elva Serna, Ninfa Vera de Bilbao, Luis Sanabria, Gloria Yaluff, Héctor Nakayama, Antonieta Rojas de Arias, Celeste Vega, Miriam Rolan, Alicia Gómez-Barrio, Hugo Cerecetto, Mercedes Gonzalez
ABSTRACT
Three series of benzimidazole N-oxide derivatives were developed and were examined for their activity against trypanosomatid parasites (Trypanosoma cruzi and Leishmania spp.). 2H-benzimidazole 1,3-dioxides displayed remarkable in vitro activities against both parasites, with derivatives 28, 29, and 32 being the most potent (IC50 < 5 microM) against the epimastigote form of T. cruzi and 28, 33, and 35 the most potent against the promastigote form of Leishmania spp. Unspecific cytotoxicity was evaluated using murine macrophages, and derivative 33 was not toxic at a concentration 30 times that of its IC50 against T. cruzi that was completely toxic for Leishmania spp., implying that the series of 2H-benzimidazole 1,3-dioxides is selective toward both trypanosomatid parasites. Derivatives 33 and 35 were submitted to an in vivo assay using an acute model of Chagas' disease and a short-term treatment (30 mg/kg/day orally administrated as aqueous solution, during 10 days). While in the control (untreated) and Benznidazole (50 mg/kg/day) groups survival fraction was 60.0% and 87.5%, respectively, none of the animals treated with derivatives 33 and 35 died. From the preliminary structure-activity relationship studies reduction potential and electrophilicity were found relevant to anti-T. cruzi activity. Active compounds are better electrophiles and more easily reduced than inactive ones.
Sigma-Aldrich
Crystal Violet, certified by the BSC
Sigma-Aldrich
Crystal Violet, S. No.: 785
Supelco
Crystal Violet, VETRANAL®, analytical standard
Sigma-Aldrich
Gentian violet
Sigma-Aldrich
Crystal Violet
Sigma-Aldrich
Crystal Violet, ACS reagent, ≥90.0% anhydrous basis
Sigma-Aldrich
Gentian Violet, meets USP testing specifications