Merck
  • Repeat hepatic resection VS radiofrequency ablation for the treatment of recurrent hepatocellular carcinoma: an updated meta-analysis.
Repeat hepatic resection VS radiofrequency ablation for the treatment of recurrent hepatocellular carcinoma: an updated meta-analysis.
Minimally invasive therapy & allied technologies : MITAT : official journal of the Society for Minimally Invasive Therapy (2020-11-05)
Junjie Liu, Jinming Zhao, H A O Gu, Zhiqiang Zhu
ABSTRACT
This paper evaluates the efficacy and safety of repeat hepatic resection and radiofrequency ablation in the treatment of recurrent hepatocellular carcinoma. We retrieved and collected all relevant articles from the inception to 8 March 2020. After data extraction, we conducted meta-analysis and carried out the heterogeneity test, sensitivity analysis, and publication bias test to evaluate reliability. A total of 12 studies with 1746 patients (rHR 837, RFA 909) were included. rHR was similar to RFA in a one-year overall survival rate (OS), while rHR was superior to RFA in 3- and 5-year OS and 1-, 3-, and 5-year disease-free survival rates (DFS), but the procedure-related complications of RFA were significantly less than those of rHR. Among the subgroups with Milan criteria, rHR was similar to RFA in 1-, 3-, and 5-year OS and 1-year DFS, but superior to RFA in 3- and 5-year DFS. RFA is the first choice for recurrent HCC meeting Milan criteria. When it does not meet the Milan criteria, minimally invasive treatment should not be carried out at the cost of survival, and rHR should be the first choice.
Sigma-Aldrich
BocA1V2PF1-NHPEG3-NH2, ≥85%
Pricing and availability is not currently available.