Merck
A labdane diterpene glucoside from the rhizomes of Curcuma mangga.
Journal of natural products (2005-07-26)
Faridah Abas, Nordin H Lajis, Khozirah Shaari, Daud Ahmad Israf, Johnson Stanslas, Umi Kalsom Yusuf, Salahuddin Mohd Raof
ABSTRACT
A new labdane diterpene glucoside, curcumanggoside (1), together with nine known compounds, including labda-8(17),12-diene-15,16-dial (2), calcaratarin A (3), zerumin B (4), scopoletin, demethoxycurcumin, bisdemethoxycurcumin, 1,7-bis(4-hydroxyphenyl)-1,4,6-heptatrien-3-one, curcumin, and p-hydroxycinnamic acid, have been isolated from the rhizomes of Curcuma mangga. Their structures were determined using a combination of 1D (1H NMR, 13C NMR, DEPT) and 2D (COSY, HSQC, HMBC) NMR techniques. All diarylheptanoids and scopoletin showed significant antioxidant activity. Zerumin B, demethoxycurcumin, bisdemethoxycurcumin, and curcumin also exhibited cytotoxic activity against a panel of five human tumor cell lines.
Product Description
Supelco
3,5-Di-tert-butyl-4-hydroxytoluene, analytical standard
Sigma-Aldrich
2,6-Di-tert-butyl-4-methylphenol, tested according to Ph. Eur.
Sigma-Aldrich
2,6-Di-tert-butyl-4-methylphenol, purum, ≥99.0% (GC)
Sigma-Aldrich
(+)-α-Tocopherol, from vegetable oil, Type V, ~1000 IU/g
Sigma-Aldrich
(+)-α-Tocopherol, Type VI, from vegetable oil, liquid (≥0.88M based on potency, density and molecular wt.), BioReagent, suitable for insect cell culture, ≥1000 IU/g
Sigma-Aldrich
Scopoletin, ≥99%
Supelco
Bisdemethoxycurcumin, analytical standard
Sigma-Aldrich
Butylated hydroxytoluene, ≥99%, FCC, FG