Merck
  • CD147-spike protein is a novel route for SARS-CoV-2 infection to host cells.
CD147-spike protein is a novel route for SARS-CoV-2 infection to host cells.
Signal transduction and targeted therapy (2020-12-06)
Ke Wang, Wei Chen, Zheng Zhang, Yongqiang Deng, Jian-Qi Lian, Peng Du, Ding Wei, Yang Zhang, Xiu-Xuan Sun, Li Gong, Xu Yang, Lei He, Lei Zhang, Zhiwei Yang, Jie-Jie Geng, Ruo Chen, Hai Zhang, Bin Wang, Yu-Meng Zhu, Gang Nan, Jian-Li Jiang, Ling Li, Jiao Wu, Peng Lin, Wan Huang, Liangzhi Xie, Zhao-Hui Zheng, Kui Zhang, Jin-Lin Miao, Hong-Yong Cui, Min Huang, Jun Zhang, Ling Fu, Xiang-Min Yang, Zhongpeng Zhao, Shihui Sun, Hongjing Gu, Zhe Wang, Chun-Fu Wang, Yacheng Lu, Ying-Ying Liu, Qing-Yi Wang, Huijie Bian, Ping Zhu, Zhi-Nan Chen
ABSTRACT
In face of the everlasting battle toward COVID-19 and the rapid evolution of SARS-CoV-2, no specific and effective drugs for treating this disease have been reported until today. Angiotensin-converting enzyme 2 (ACE2), a receptor of SARS-CoV-2, mediates the virus infection by binding to spike protein. Although ACE2 is expressed in the lung, kidney, and intestine, its expressing levels are rather low, especially in the lung. Considering the great infectivity of COVID-19, we speculate that SARS-CoV-2 may depend on other routes to facilitate its infection. Here, we first discover an interaction between host cell receptor CD147 and SARS-CoV-2 spike protein. The loss of CD147 or blocking CD147 in Vero E6 and BEAS-2B cell lines by anti-CD147 antibody, Meplazumab, inhibits SARS-CoV-2 amplification. Expression of human CD147 allows virus entry into non-susceptible BHK-21 cells, which can be neutralized by CD147 extracellular fragment. Viral loads are detectable in the lungs of human CD147 (hCD147) mice infected with SARS-CoV-2, but not in those of virus-infected wild type mice. Interestingly, virions are observed in lymphocytes of lung tissue from a COVID-19 patient. Human T cells with a property of ACE2 natural deficiency can be infected with SARS-CoV-2 pseudovirus in a dose-dependent manner, which is specifically inhibited by Meplazumab. Furthermore, CD147 mediates virus entering host cells by endocytosis. Together, our study reveals a novel virus entry route, CD147-spike protein, which provides an important target for developing specific and effective drug against COVID-19.