Merck
  • Antibodies against Phosphorylcholine and Malondialdehyde during the First Two Years of Life.
Antibodies against Phosphorylcholine and Malondialdehyde during the First Two Years of Life.
Journal of immunology (Baltimore, Md. : 1950) (2020-09-06)
Divya Thiagarajan, Susanna L Lundström, Göran Pershagen, Catarina Almqvist Malmros, Ellika Andolf, Anna Hedman, Oscar Berg, Nina Oparina, Johan Frostegård
ABSTRACT
Abs against phosphorylcholine (anti-PC) and Abs against malondialdehyde (anti-MDA) may be protective in chronic inflammation, like atherosclerosis and cardiovascular disease. It is not known how they develop early in life. Ab titers were measured using ELISA in healthy women (n = 105; born into life study) and their children. Plasma samples were collected from the mothers before conception and from the children at birth as well as at 1 and 2 y after birth. Extracted Abs were compared using a proteomics de novo sequencing approach. It was observed that children were born with very low levels of IgM anti-PC, whereas IgM anti-MDA was present at birth. Both IgM anti-PC and anti-MDA increased during the first 2 y of life, but IgM anti-PC in contrast to IgM anti-MDA was still significantly lower than in the mothers. IgG anti-PC decreased after 1 y but reached similar levels as mothers' after 2 y, whereas IgG anti-MDA reached similar levels as mothers' already after 1 y. Proteomics peptide sequencing analysis indicated large peptide sequence variation without specific clone expression during the early stage of life compared with the adult stage for which specific peptide sequences dominated. IgM anti-PC levels develop much slower than anti-MDA and are still relatively low at 2 y. We hypothesize that anti-PC is developed by a combination of preprogramming and exposure to the external world, in which infectious agents may play a role. For anti-MDA, preprogramming is likely to play a major role and at an earlier stage than for anti-PC.
Sigma-Aldrich
Anti-Human IgG (Fc specific)−Biotin antibody, Mouse monoclonal, clone HP-6017, purified from hybridoma cell culture