Merck
  • SATB2 in neuroendocrine neoplasms: strong expression is restricted to well-differentiated tumours of lower gastrointestinal tract origin and is most frequent in Merkel cell carcinoma among poorly differentiated carcinomas.
SATB2 in neuroendocrine neoplasms: strong expression is restricted to well-differentiated tumours of lower gastrointestinal tract origin and is most frequent in Merkel cell carcinoma among poorly differentiated carcinomas.
Histopathology (2019-06-25)
Andrew M Bellizzi
ABSTRACT
Special AT-rich sequence-binding protein 2 (SATB2) is a transcriptional regulator with critical roles in brain, craniofacial and skeletal development. It has emerged as a key marker of lower gastrointestinal (GI) tract columnar epithelial and osteoblastic differentiation. Transcription factor immunohistochemistry is useful in assigning site of origin in well-differentiated neuroendocrine tumours (NETs), and has had a limited role in poorly differentiated neuroendocrine carcinomas (NECs). This study sought to evaluate the role of SATB2 in assigning site of origin in neuroendocrine epithelial neoplasms. Tissue microarrays were constructed from the following: 317 NETs (37 thyroid, 46 lung, 16 stomach, 12 duodenum, 70 pancreas, 106 jejunoileum, 24 appendix, and six rectosigmoid), 44 phaeochromocytomas/paragangliomas, and 79 NECs (29 Merkel cell, 30 lung, and 20 extrapulmonary visceral); nine appendiceal and 19 rectal NETs were examined in whole sections. SATB2 immunohistochemistry was scored for extent (%) and intensity (0-3+), with an H-score being calculated. SATB2 was expressed by 96% of rectosigmoid NETs, 79% of appendiceal NETs, and only 7% of other well-differentiated neoplasms (P < 0.0001). Expression in lower GI tract NETs (median H-score of 255) was stronger than in other positive tumours (median H-score of 7) (P < 0.0001). Any SATB2 expression was 86% sensitive/93% specific for lower GI tract origin. SATB2 was expressed by 79% of Merkel cell carcinomas (median H-score of 300), 33% of lung NECs (median H-score of 23), and 60% of extrapulmonary visceral NECs (median H-score of 110), with stronger expression in Merkel cell carcinoma (P < 0.001). At an H-score cutoff of ≥150, SATB2 was 69% sensitive/90% specific for Merkel cell carcinoma. SATB2 is frequently and strongly expressed by lower GI tract NETs; we have adopted it as our rectal NET marker. Relatively frequent and strong expression in Merkel cell carcinoma may have value in assigning NEC site of origin.