Merck
  • Low band gap dithieno[3,2-b:2',3'-d]silole-containing polymers, synthesis, characterization and photovoltaic application.
Low band gap dithieno[3,2-b:2',3'-d]silole-containing polymers, synthesis, characterization and photovoltaic application.
Chemical communications (Cambridge, England) (2009-09-16)
Lijun Huo, Hsiang-Yu Chen, Jianhui Hou, Teresa L Chen, Yang Yang
ABSTRACT
A series of low band gap silole-containing polymers were synthesized with different alkyl side chains and a power conversion efficiency (PCE) of 3.43% was obtained.