Merck
  • Thermus thermophilus Argonaute Functions in the Completion of DNA Replication.
Thermus thermophilus Argonaute Functions in the Completion of DNA Replication.
Cell (2020-08-28)
Samson M Jolly, Ildar Gainetdinov, Karina Jouravleva, Han Zhang, Lara Strittmatter, Shannon M Bailey, Gregory M Hendricks, Avantika Dhabaria, Beatrix Ueberheide, Phillip D Zamore
ABSTRACT
In many eukaryotes, Argonaute proteins, guided by short RNA sequences, defend cells against transposons and viruses. In the eubacterium Thermus thermophilus, the DNA-guided Argonaute TtAgo defends against transformation by DNA plasmids. Here, we report that TtAgo also participates in DNA replication. In vivo, TtAgo binds 15- to 18-nt DNA guides derived from the chromosomal region where replication terminates and associates with proteins known to act in DNA replication. When gyrase, the sole T. thermophilus type II topoisomerase, is inhibited, TtAgo allows the bacterium to finish replicating its circular genome. In contrast, loss of gyrase and TtAgo activity slows growth and produces long sausage-like filaments in which the individual bacteria are linked by DNA. Finally, wild-type T. thermophilus outcompetes an otherwise isogenic strain lacking TtAgo. We propose that the primary role of TtAgo is to help T. thermophilus disentangle the catenated circular chromosomes generated by DNA replication.
Sigma-Aldrich
Cyanogen bromide-activated-Sepharose 4B, lyophilized powder
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Protocatechuate 3,4-Dioxygenase from Pseudomonas sp., lyophilized powder, ≥3 units/mg solid
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
D.E.R. 332, used as embedding medium
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
4-Nitrobenzyl alcohol, 99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
3,4-Dihydroxybenzoic acid, ≥97.0% (T)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
(±)-6-Hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid, 97%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Propyl gallate, powder