Merck
  • Electron ionization induced fragmentation of fluorinated derivatives of bisphenols.
Electron ionization induced fragmentation of fluorinated derivatives of bisphenols.
Rapid communications in mass spectrometry : RCM (2020-06-14)
Magda Caban, Piotr Stepnowski
ABSTRACT
Fluorinated derivatization agents allow for the identification and quantification of emerging pollutants with high sensitivity, yet details of their potential applications using electron ionization are lacking. The fluorine atom itself does not effectively participate in electron ionization. Furthermore, limited information exists regarding the effect of fluorine during electron ionization-induced fragmentation. To fill this gap, this report presents the fragmentation pathways of the fluorinated derivatives of ten bisphenol analogues as exemplary emerging pollutants. The bisphenols were derivatized by the acetylation reagent trifluoroacetic anhydride and a new silylation reagent, namely dimethyl(3,3,3-trifluoropropyl)silyldiethylamine (DIMETRIS; previously applied for the analysis of selected pharmaceuticals in environmental samples), and analyzed using GC/MS (electron ionization, 70 eV). Deuterated bisphenol A was added to the group of analytes to confirm the proposed fragmentation pathways. The specific chemical structure of bisphenols gives the possibility of several resonance hybrids of C-centered radicals. This, in turn, results in several fragmentation pathways, unique for each resonance hybrid. Sequential losses of radicals and neutral fragments were observed in both types of derivative, with final stable carbenium ions. McLafferty-type rearrangements were observed between the native structure of the analytes and the introduced substituents. The gamma-shift of F onto Si in the Si(CH2 )2 CF3 substituent is proposed to explain the loss of the fragment with a mass of 78 u. Both types of derivatization reagent used were found to be applicable, although the use of DIMETRIS was limited for high-mass bisphenols. The introduction of fluorine by derivatization brings benefits for the qualitative and quantitative analysis of bisphenol-type compounds using GC/MS because of the presence of characteristic ions in the mass spectra.
Sigma-Aldrich
4,4′-Cyclohexylidenebisphenol, 98%
Sigma-Aldrich
4,4′-(Hexafluoroisopropylidene)diphenol, 97%
Sigma-Aldrich
Bisphenol A diacetate, 98%
Sigma-Aldrich
Bis(4-hydroxyphenyl)methane, 98%
Sigma-Aldrich
Bisphenol A dimethacrylate, >98%