Merck
  • Large-Scale Topological Changes Restrain Malignant Progression in Colorectal Cancer.
Large-Scale Topological Changes Restrain Malignant Progression in Colorectal Cancer.
Cell (2020-08-26)
Sarah E Johnstone, Alejandro Reyes, Yifeng Qi, Carmen Adriaens, Esmat Hegazi, Karin Pelka, Jonathan H Chen, Luli S Zou, Yotam Drier, Vivian Hecht, Noam Shoresh, Martin K Selig, Caleb A Lareau, Sowmya Iyer, Son C Nguyen, Eric F Joyce, Nir Hacohen, Rafael A Irizarry, Bin Zhang, Martin J Aryee, Bradley E Bernstein
ABSTRACT
Widespread changes to DNA methylation and chromatin are well documented in cancer, but the fate of higher-order chromosomal structure remains obscure. Here we integrated topological maps for colon tumors and normal colons with epigenetic, transcriptional, and imaging data to characterize alterations to chromatin loops, topologically associated domains, and large-scale compartments. We found that spatial partitioning of the open and closed genome compartments is profoundly compromised in tumors. This reorganization is accompanied by compartment-specific hypomethylation and chromatin changes. Additionally, we identify a compartment at the interface between the canonical A and B compartments that is reorganized in tumors. Remarkably, similar shifts were evident in non-malignant cells that have accumulated excess divisions. Our analyses suggest that these topological changes repress stemness and invasion programs while inducing anti-tumor immunity genes and may therefore restrain malignant progression. Our findings call into question the conventional view that tumor-associated epigenomic alterations are primarily oncogenic.
Sigma-Aldrich
Triethanolamine, ≥99.0% (GC)
Sigma-Aldrich
Acetic anhydride, ReagentPlus®, ≥99%
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
3′-Sialyl-Lewis-X tetrasaccharide
Sigma-Aldrich
Dextran sulfate sodium salt from Leuconostoc spp., average Mw >500,000 (dextran starting material), contains 0.5-2% phosphate buffer
Sigma-Aldrich
Formamide, BioReagent, ≥99.5% (GC)
Sigma-Aldrich
Formamide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Ammonium chloride, ACS reagent, ≥99.5%