Merck
  • Quantitative evaluation of infectivity change of Cryptosporidium parvum after gamma irradiation.
Quantitative evaluation of infectivity change of Cryptosporidium parvum after gamma irradiation.
The Korean journal of parasitology (2009-03-18)
Soo-Ung Lee, Mikyo Joung, Taekyoung Nam, Woo-Yoon Park, Jae-Ran Yu
ABSTRACT
Cryptosporidium parvum is a well-known waterborne and opportunistic intracellular protozoan parasite that causes diarrheal illness. In this study, we quantitatively investigated reduction of the infectivity of C. parvum after gamma irradiation and repair of the infectivity during incubation time after irradiation. C. parvum oocysts were subjected to gamma irradiation at various doses (1, 5, 10, and 25 kGy), and the in vitro infectivity was measured by real-time PCR every day up to 7 days after irradiation. The in vitro infectivity of C. parvum on human ileocecal adenocarcinoma cells (HCT-8) was effectively reduced (> 2 log(10)) by irradiation at 10 kGy or more. However, in the experiment to find out repair of the infectivity, recovery was not noted until day 7 post-incubation.
Product Description
SAFC
Glasgow′s Modified Eagle′s Medium
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium, Without glucose, L-glutamine, phenol red, sodium pyruvate and sodium bicarbonate, powder, suitable for cell culture
SAFC
L-Glutamine
Sigma-Aldrich
Sodium taurocholate hydrate, ≥97.0% (TLC)
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland