Merck
  • Salivary extracellular vesicles inhibit Zika virus but not SARS-CoV-2 infection.
Salivary extracellular vesicles inhibit Zika virus but not SARS-CoV-2 infection.
Journal of extracellular vesicles (2020-09-18)
Carina Conzelmann, Rüdiger Groß, Min Zou, Franziska Krüger, André Görgens, Manuela O Gustafsson, Samir El Andaloussi, Jan Münch, Janis A Müller
ABSTRACT
Zika virus (ZIKV) is mainly transmitted via mosquitos, but human-to-human transmissions also occur. The virus is shed into various body fluids including saliva, which represents a possible source of viral transmission. Thus, we here explored whether human saliva affects ZIKV infectivity. We found that physiological concentrations of pooled saliva dose-dependently inhibit ZIKV infection of monkey and human cells by preventing viral attachment to target cells. The anti-ZIKV activity in saliva could not be abrogated by boiling, suggesting the antiviral factor is not a protein. Instead, we found that purified extracellular vesicles (EVs) from saliva inhibit ZIKV infection. Salivary EVs (saEVs) express typical EV markers such as tetraspanins CD9, CD63 and CD81 and prevent ZIKV attachment to and infection of target cells at concentrations that are naturally present in saliva. The anti-ZIKV activity of saliva is conserved but the magnitude of inhibition varies between individual donors. In contrast to ZIKV, severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2), predominantly spreading via respiratory droplets, is not affected by saliva or saEVs. Our findings provide a plausible explanation for why ZIKV transmission via saliva, i.e. by deep kissing have not been recorded and establish a novel oral innate immune defence mechanism against some viral pathogens.
Sigma-Aldrich
1,2-Dioleoyl-sn-glycero-3-phosphocholine, lyophilized powder
Sigma-Aldrich
Nitrilotriacetic acid disodium salt, Sigma Grade, ≥99%
Sigma-Aldrich
MEM Non-essential Amino Acid Solution (100×), without L-glutamine, liquid, sterile-filtered, BioReagent, suitable for cell culture
Millipore
Benzonase® Nuclease, ≥250 units/μL, ≥90% (SDS-PAGE), recombinant, expressed in E. coli, buffered aqueous glycerol solution
Vivaspin 20 centrifugal concentrator, MWCO 100 kDa