Merck
  • Activation of LXRβ Signaling in the Amygdala Confers Anxiolytic Effects Through Rebalancing Excitatory and Inhibitory Neurotransmission upon Acute Stress.
Activation of LXRβ Signaling in the Amygdala Confers Anxiolytic Effects Through Rebalancing Excitatory and Inhibitory Neurotransmission upon Acute Stress.
Neurotherapeutics : the journal of the American Society for Experimental NeuroTherapeutics (2020-04-17)
Wen Yu, Lu Wang, Le Yang, Yan-Jiao Li, Min Wang, Chen Qiu, Qi Yang, Xu-Bo Li, Yun-Long Huang, Rui Liu, Yu-Mei Wu
ABSTRACT
The balance of major excitatory (glutamate, Glu) and inhibitory (γ-aminobutyric acid, GABA), named as E/I neurotransmission, is critical for proper information processing. Anxiety-like responses upon stress are accompanied by abnormal alterations in the formation and function of synapses, resulting in the imbalance of E/I neurotransmission in the amygdala. Liver X receptors (LXRs), including LXRα and LXRβ isoforms, are nuclear receptors responsible for regulating central nervous system (CNS) functions besides maintaining metabolic homeostasis. However, little is known about the contribution of LXRs in E/I balance in regulating anxiety-related behaviors induced by stress. In this study, we found stress-induced anxiety led to the expression reduction of LXRβ not LXRα in mice amygdala. GW3965, a dual agonist for both LXRα and LXRβ, alleviated anxiety-like behaviors of stressed mice through activation of LXRβ, confirmed by the knockdown of LXRβ mediated by lentiviral shRNAs in the basolateral amygdala (BLA). This was paralleled by correcting the disequilibrium of E/I neurotransmission in the stressed BLA. Importantly, GW3965 exerted anxiolytic effects by correcting the promoted amplitude and frequency of miniature excitatory postsynaptic current (mEPSC), and augmenting the decreased that of miniature inhibitory postsynaptic current (mIPSC) in the stressed BLA. This suggests that stress-induced anxiety-like behaviors can largely be ascribed to the deficit of LXRβ signaling in E/I neurotransmission in BLA. These findings highlight the deficiency of LXRβ signaling in the amygdala linked to anxiety disorder, and LXRβ activation may represent a potential novel target for anxiety treatment with an alteration in synaptic transmission in the amygdala.
Sigma-Aldrich
Anti-NMDAR2A Antibody, Chemicon®, from mouse
Supelco
γ-Aminobutyric acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Anti-NMDAR2B Antibody, ascites fluid, clone 1E4.25A4, Chemicon®
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
L-Glutamic acid monosodium salt monohydrate, ≥98.0% (NT)
Sigma-Aldrich
1-Fluoro-2,4-dinitrobenzene, purum p.a., ≥98.0% (GC)