Merck
  • The novel ciliogenesis regulator DYRK2 governs Hedgehog signaling during mouse embryogenesis.
The novel ciliogenesis regulator DYRK2 governs Hedgehog signaling during mouse embryogenesis.
eLife (2020-08-08)
Saishu Yoshida, Katsuhiko Aoki, Ken Fujiwara, Takashi Nakakura, Akira Kawamura, Kohji Yamada, Masaya Ono, Satomi Yogosawa, Kiyotsugu Yoshida
ABSTRACT
Mammalian Hedgehog (Hh) signaling plays key roles in embryogenesis and uniquely requires primary cilia. Functional analyses of several ciliogenesis-related genes led to the discovery of the developmental diseases known as ciliopathies. Hence, identification of mammalian factors that regulate ciliogenesis can provide insight into the molecular mechanisms of embryogenesis and ciliopathy. Here, we demonstrate that DYRK2 acts as a novel mammalian ciliogenesis-related protein kinase. Loss of Dyrk2 in mice causes suppression of Hh signaling and results in skeletal abnormalities during in vivo embryogenesis. Deletion of Dyrk2 induces abnormal ciliary morphology and trafficking of Hh pathway components. Mechanistically, transcriptome analyses demonstrate down-regulation of Aurka and other disassembly genes following Dyrk2 deletion. Taken together, the present study demonstrates for the first time that DYRK2 controls ciliogenesis and is necessary for Hh signaling during mammalian development.
Sigma-Aldrich
MISSION® esiRNA, targeting human DYRK2
Sigma-Aldrich
MISSION® esiRNA, targeting human UBE2C
Sigma-Aldrich
Smoothened Agonist, SAG, A cell-permeable Smoothened Agonist, SAG, CAS 364590-63-6, modulates the coupling of Smo with its downstream effector by interacting with the Smo heptahelical domain (KD = 59 nM).
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Dyrk2
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Aurka
Sigma-Aldrich
Anti-Acetylated Tubulin antibody, Mouse monoclonal, clone 6-11B-1, purified from hybridoma cell culture