Merck
  • Colonisation with extended spectrum beta-lactamase-producing and carbapenem-resistant Enterobacterales in children admitted to a paediatric referral hospital in South Africa.
Colonisation with extended spectrum beta-lactamase-producing and carbapenem-resistant Enterobacterales in children admitted to a paediatric referral hospital in South Africa.
PloS one (2020-11-07)
Babatunde O Ogunbosi, Clinton Moodley, Preneshni Naicker, James Nuttall, Colleen Bamford, Brian Eley
ABSTRACT
There are few studies describing colonisation with extended spectrum beta-lactamase-producing Enterobacterales (ESBL-PE) and carbapenem-resistant Enterobacterales (CRE) among children in sub-Saharan Africa. Colonisation often precedes infection and multi-drug-resistant Enterobacterales are important causes of invasive infection. In this prospective cross-sectional study, conducted between April and June 2017, 200 children in a tertiary academic hospital were screened by rectal swab for EBSL-PE and CRE. The resistance-conferring genes were identified using polymerase chain reaction technology. Risk factors for colonisation were also evaluated. Overall, 48% (96/200) of the children were colonised with at least one ESBL-PE, 8.3% (8/96) of these with 2 ESBL-PE, and one other child was colonised with a CRE (0.5% (1/200)). Common colonising ESBL-PE were Klebsiella pneumoniae (62.5%, 65/104) and Escherichia coli (34.6%, 36/104). The most frequent ESBL-conferring gene was blaCTX-M in 95% (76/80) of the isolates. No resistance- conferring gene was identified in the CRE isolate (Enterobacter cloacae). Most of the Klebsiella pneumoniae isolates were susceptible to piperacillin/tazobactam (86.2%) and amikacin (63.9%). Similarly, 94.4% and 97.2% of the Escherichia coli isolates were susceptible to piperacillin/tazobactam and amikacin, respectively. Hospitalisation for more than 7 days before study enrolment was associated with ESBL-PE colonisation. Approximately half of the hospitalised children in this study were colonised with ESBL-PE. This highlights the need for improved infection prevention and control practices to limit the dissemination of these microorganisms.
Product Description
Supelco
Imipenem, Pharmaceutical Secondary Standard; Certified Reference Material
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Estimated to ship on March 21, 2025
$105.00
1 amp
Estimated to ship on March 21, 2025
$117.00
25 g
Estimated to ship on March 21, 2025
$468.00
10 amp
Estimated to ship on March 21, 2025
$472.00
100 g
Estimated to ship on March 21, 2025
$1,360.00