Merck
  • Deubiquitinase USP20 promotes breast cancer metastasis by stabilizing SNAI2.
Deubiquitinase USP20 promotes breast cancer metastasis by stabilizing SNAI2.
Genes & development (2020-09-19)
Wenyang Li, Minhong Shen, Yi-Zhou Jiang, Ruina Zhang, Hanqiu Zheng, Yong Wei, Zhi-Ming Shao, Yibin Kang
ABSTRACT
SNAI2/SLUG, a metastasis-promoting transcription factor, is a labile protein that is degraded through the ubiquitin proteasome degradation system. Here, we conducted comprehensive gain- and loss-of-function screens using a human DUB cDNA library of 65 genes and an siRNA library of 98 genes, and identified USP20 as a deubiquitinase (DUB) that regulates SNAI2 ubiquitination and stability. Further investigation of USP20 demonstrated its function in promoting migration, invasion, and metastasis of breast cancer. USP20 positively correlates with SNAI2 protein level in breast tumor samples, and higher USP20 expression is associated with poor prognosis in ER- breast cancer patients.
Product Description
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial
Sigma-Aldrich
MISSION® esiRNA, targeting human SNAI2
Sigma-Aldrich
MISSION® esiRNA, targeting human USP20