Merck
  • Sirtuin 7 promotes non‑small cell lung cancer progression by facilitating G1/S phase and epithelial‑mesenchymal transition and activating AKT and ERK1/2 signaling.
Sirtuin 7 promotes non‑small cell lung cancer progression by facilitating G1/S phase and epithelial‑mesenchymal transition and activating AKT and ERK1/2 signaling.
Oncology reports (2020-07-25)
Yingying Zhao, Xia Ye, Ruifang Chen, Qian Gao, Daguo Zhao, Chunhua Ling, Yulan Qian, Chun Xu, Min Tao, Yufeng Xie
ABSTRACT
Increasing evidence has indicated the roles of sirtuin 7 (SIRT7) in numerous human cancers. However, the effects and the clinical significance of SIRT7 in human lung cancer is largely unknown. The present research demonstrated that SIRT7 was increased in human lung cancer tumor tissues. SIRT7 upregulation was associated with clinicopathological characteristics of lung cancer malignancy including positive lymph node metastasis, high pathologic stage and large tumor size. SIRT7 was also upregulated in human non‑small cell lung cancer (NSCLC) cell lines. Furthermore SIRT7‑overexpressed A549 (A549‑SIRT7) and SIRT7‑knocked down H292 (H292‑shSIRT7) human NSCLC cell lines were established. Using these NSCLC cells and xenograft mouse models, it was revealed that SIRT7 overexpression markedly promoted growth and G1 to S cell cycle phase transition as well as migration, invasion and distant lung metastasis in A549 NSCLC cells, whereas SIRT7 knockdown suppressed these processes in H292 NSCLC cells. Mechanistically, in A549 NSCLC cells, SIRT7 overexpression significantly activated not only protein kinase B (AKT) signaling but also extracellular signal‑regulated kinase 1/2 (ERK1/2) signaling. SIRT7 overexpression also significantly downregulated cyclin‑dependent kinase (CDK) inhibitors including p21 and p27 as well as upregulated cyclins including cyclin D1 and cyclin E1, and CDKs including CDK2 and CDK4. Notably, the epithelial‑mesenchymal transition (EMT) process of A549 NSCLC cells was facilitated by SIRT7 overexpression, as evidenced by E‑cadherin epithelial marker downregulation and mesenchymal markers (N‑cadherin, vimentin, Snail and Slug) upregulation. In addition, SIRT7 knockdown in H292 NSCLC cells exhibited the opposite regulatory effects. Moreover, inhibition of AKT signaling abated the promoting effects of SIRT7 in NSCLC cell proliferation and EMT progression. The present data indicated that SIRT7 accelerated human NSCLC cell growth and metastasis possibly by promotion of G1 to S‑phase transition and EMT through modulation of the expression of G1‑phase checkpoint molecules and EMT markers as well as activation of AKT and ERK1/2 signaling. SIRT7 could be an innovative potential target for human NSCLC therapy.
Sigma-Aldrich
MISSION® esiRNA, targeting human SIRT7
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder