Merck
  • ACSL3 is a novel GABARAPL2 interactor that links ufmylation and lipid droplet biogenesis.
ACSL3 is a novel GABARAPL2 interactor that links ufmylation and lipid droplet biogenesis.
Journal of cell science (2020-08-28)
Franziska Eck, Santosh Phuyal, Matthew D Smith, Manuel Kaulich, Simon Wilkinson, Hesso Farhan, Christian Behrends
ABSTRACT
While studies of the autophagy-related (ATG) genes in knockout models have led to an explosion of knowledge about the functions of autophagy components, the exact roles of LC3 and GABARAP family proteins (human ATG8 equivalents) are still poorly understood. A major drawback in understanding their roles is that the available interactome data has largely been acquired using overexpression systems. To overcome these limitations, we employed CRISPR/Cas9-based genome-editing to generate a panel of cells in which human ATG8 genes were tagged at their natural chromosomal locations with an N-terminal affinity epitope. This cellular resource was employed to map endogenous GABARAPL2 protein complexes using interaction proteomics. This approach identified the ER-associated protein and lipid droplet (LD) biogenesis factor ACSL3 as a stabilizing GABARAPL2-binding partner. GABARAPL2 bound ACSL3 in a manner dependent on its LC3-interacting regions, whose binding site in GABARAPL2 was required to recruit the latter to the ER. Through this interaction, the UFM1-activating enzyme UBA5 became anchored at the ER. Furthermore, ACSL3 depletion and LD induction affected the abundance of several ufmylation components and ER-phagy. Together these data allow us to define ACSL3 as a novel regulator of the enigmatic UFM1 conjugation pathway.
Sigma-Aldrich
Endoplasmic Reticulum Isolation Kit, isolation of intact ER from mammalian soft tissues and cultured cells
Sigma-Aldrich
Monoclonal Anti-HA antibody produced in mouse, clone HA-7, ascites fluid
Sigma-Aldrich
Earle′s Balanced Salts, With sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Doxycycline hyclate