Merck
  • Quantification of F2-isoprostanes as a reliable index of oxidative stress in vivo using gas chromatography-mass spectrometry (GC-MS) method.
Quantification of F2-isoprostanes as a reliable index of oxidative stress in vivo using gas chromatography-mass spectrometry (GC-MS) method.
Free radical biology & medicine (2009-08-04)
Wei Liu, Jason D Morrow, Huiyong Yin
ABSTRACT
Free radical-induced lipid peroxidation has been implicated in a number of human diseases including atherosclerosis, cancer, and neurodegenerative diseases. F(2)-Isoprostanes (IsoPs) are isomers of prostaglandin PGF(2alpha) that are generated in vivo from the free radical-initiated peroxidation of arachidonic acid independent of cyclooxygenase enzymes. Since the discovery of the IsoPs in the early 1990s, a large body of evidence has been accumulated to indicate that quantification of these F(2)-IsoPs represents the most reliable biomarker to assess oxidative stress in vivo. A variety of analytical approaches have been developed for the quantification of these novel compounds; these methods include mass spectrometry (MS) detection coupled to gas chromatography (GC) or liquid chromatography (LC) separation, and detection using immunological approaches. This article summarizes our current methodology to quantify F(2)-IsoPs in biological fluids and tissues using GC-MS. This method includes solid-phase extraction (SPE), thin-layer chromatography (TLC) purification, chemical derivatization, and MS detection using negative ion chemical ionization (NICI) coupled with GC. The protocol described herein has been optimized and validated to provide the best sensitivity and selectivity for quantification of F(2)-IsoPs from a variety of biological sources.
Sigma-Aldrich
Butylated hydroxytoluene, ≥99%, FCC, FG
Sigma-Aldrich
Undecane, ≥99%
Sigma-Aldrich
Phosphomolybdic acid solution, ready-to-use spray and plunge reagent, for chromatography
Supelco
Undecane, analytical standard
Sigma-Aldrich
Phosphomolybdic acid solution, spray reagent, 10% in ethanol
Sigma-Aldrich
Phosphomolybdic acid solution, 20 wt. % in ethanol
Sigma-Aldrich
Undecane, ≥99%
Supelco
Butylated Hydroxytoluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Triphenylphosphine, ReagentPlus®, 99%
Sigma-Aldrich
Triphenylphosphine, ≥95.0% (GC)