Merck
  • Leukotriene E4-induced pulmonary inflammation is mediated by the P2Y12 receptor.
Leukotriene E4-induced pulmonary inflammation is mediated by the P2Y12 receptor.
The Journal of experimental medicine (2009-10-14)
Sailaja Paruchuri, Hiroyuki Tashimo, Chunli Feng, Akiko Maekawa, Wei Xing, Yongfeng Jiang, Yoshihide Kanaoka, Pamela Conley, Joshua A Boyce
ABSTRACT
Of the potent lipid inflammatory mediators comprising the cysteinyl leukotrienes (LTs; LTC4, LTD4, and LTE4), only LTE4 is stable and abundant in vivo. Although LTE4 shows negligible activity at the type 1 and 2 receptors for cys-LTs (CysLT1R and CysLT2R), it is a powerful inducer of mucosal eosinophilia and airway hyperresponsiveness in humans with asthma. We show that the adenosine diphosphate (ADP)-reactive purinergic (P2Y12) receptor is required for LTE4-mediated pulmonary inflammation. P2Y12 receptor expression permits LTE4-induced activation of extracellular signal-regulated kinase in Chinese hamster ovary cells and permits chemokine and prostaglandin D2 production by LAD2 cells, a human mast cell line. P2Y12 receptor expression by LAD2 cells is required for competition between radiolabeled ADP and unlabeled LTE4 but not for direct binding of LTE4, suggesting that P2Y12 complexes with another receptor to recognize LTE4. Administration of LTE4 to the airways of sensitized mice potentiates eosinophilia, goblet cell metaplasia, and expression of interleukin-13 in response to low-dose aerosolized allergen. These responses persist in mice lacking both CysLT1R and CysLT2R but not in mice lacking P2Y12 receptors. The effects of LTE4 on P2Y12 in the airway were abrogated by platelet depletion. Thus, the P2Y12 receptor is required for proinflammatory actions of the stable abundant mediator LTE4 and is a novel potential therapeutic target for asthma.
Sigma-Aldrich
Apyrase from potatoes, ATPase ≥200 units/mg protein, lyophilized powder
Sigma-Aldrich
Apyrase from potatoes, High Activity, ATPase ≥600 units/mg protein, lyophilized powder
Sigma-Aldrich
Apyrase from potatoes, ATPase ≥3.0 units/mg protein, lyophilized powder
Sigma-Aldrich
Apyrase from potatoes, ATPase ≥60 units/mg protein, lyophilized powder (partially purified)
Sigma-Aldrich
Apyrase from potatoes, ATPase ≥200 units/mg protein, lyophilized powder
Sigma-Aldrich
Fetal Bovine Serum, Australia origin, γ-irradiated, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, Australia origin, USDA approved, Heat Inactivated, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Heat Inactivated, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, USA origin, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Fetal Bovine Serum, sterile-filtered, non-USA origin, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, suitable for, Australia origin, USDA approved, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Fetal Bovine Serum, Heat Inactivated, non-USA origin, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Heat Inactivated, sterile-filtered, suitable for cell culture, suitable for insect cell culture, suitable for hybridoma
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Charcoal Stripped, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, Canada origin, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, Australia origin, USDA approved, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, USA origin, suitable for cell culture