Merck
Translation and Replication Dynamics of Single RNA Viruses.
Cell (2020-11-15)
Sanne Boersma, Huib H Rabouw, Lucas J M Bruurs, Tonja Pavlovič, Arno L W van Vliet, Joep Beumer, Hans Clevers, Frank J M van Kuppeveld, Marvin E Tanenbaum
ABSTRACT
RNA viruses are among the most prevalent pathogens and are a major burden on society. Although RNA viruses have been studied extensively, little is known about the processes that occur during the first several hours of infection because of a lack of sensitive assays. Here we develop a single-molecule imaging assay, virus infection real-time imaging (VIRIM), to study translation and replication of individual RNA viruses in live cells. VIRIM uncovered a striking heterogeneity in replication dynamics between cells and revealed extensive coordination between translation and replication of single viral RNAs. Furthermore, using VIRIM, we identify the replication step of the incoming viral RNA as a major bottleneck of successful infection and identify host genes that are responsible for inhibition of early virus replication. Single-molecule imaging of virus infection is a powerful tool to study virus replication and virus-host interactions that may be broadly applicable to RNA viruses.
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Cy5-azide, 95% (HPLC)
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, for fluorescence, mixture of bleomycin sulfate salts, lyophilized, powder or crystals, white to off-white
Sigma-Aldrich
DL-Cysteine, technical grade
Sigma-Aldrich
Rupintrivir, ≥98% (HPLC)