Merck
  • Immunohistochemistry in the evaluation of neovascularization in tumor xenografts.
Immunohistochemistry in the evaluation of neovascularization in tumor xenografts.
Biotechnic & histochemistry : official publication of the Biological Stain Commission (2008-10-11)
D Wang, C R Stockard, L Harkins, P Lott, C Salih, K Yuan, D Buchsbaum, A Hashim, M Zayzafoon, R W Hardy, O Hameed, W Grizzle, G P Siegal
ABSTRACT
Angiogenesis, or neovascularization, is known to play an important role in the neoplastic progression leading to metastasis. CD31 or Factor VIII-related antigen (F VIII RAg) immunohistochemistry is widely used in experimental studies for quantifying tumor neovascularization in immunocompromised animal models implanted with transformed human cell lines. Quantification, however, can be affected by variations in the methodology used to measure vascularization including antibody selection, antigen retrieval (AR) pretreatment, and evaluation techniques. To examine this further, we investigated the microvessel density (MVD) and the intensity of microvascular staining among five different human tumor xenografts and a mouse syngeneic tumor using anti-CD31 and F VIII RAg immunohistochemical staining. Different AR methods also were evaluated. Maximal retrieval of CD31 was achieved using 0.5 M Tris (pH 10) buffer, while maximum retrieval of F VIII RAg was achieved using 0.05% pepsin treatment of tissue sections. For each optimized retrieval condition, anti-CD31 highlighted small vessels better than F VIII RAg. Furthermore, the MVD of CD31 was significantly greater than that of F VIII RAg decorated vessels (p<0.001). The choice of antibody and AR method has a significant affect on immunohistochemical findings when studying angiogenesis. One also must use caution when comparing studies in the literature that use different techniques and reagents.
Sigma-Aldrich
Pepsin from porcine gastric mucosa, tested according to Ph. Eur.
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, slightly beige, ≥500 U/mg
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, slightly beige, 1200-2400 U/mg
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, ≥250 units/mg solid
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, ≥400 units/mg protein
Sigma-Aldrich
Pepsin from porcine gastric mucosa, lyophilized powder, ≥2,500 units/mg protein (E1%/280)
Sigma-Aldrich
Pepsin from porcine gastric mucosa, lyophilized powder, ≥3,200 units/mg protein
Supelco
Biotin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Biotin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Biotin, tested according to Ph. Eur.
Sigma-Aldrich
Biotin, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Biotin, meets USP testing specifications
Sigma-Aldrich
Goat serum
Sigma-Aldrich
Biotin, ≥99% (HPLC), lyophilized powder