Merck
SIRT1 accelerates the progression of activity-based anorexia.
Nature communications (2020-06-06)
Timothy M Robinette, Justin W Nicholatos, Adam B Francisco, Kayla E Brooks, Rachel Y Diao, Sandro Sorbi, Valdo Ricca, Benedetta Nacmias, Miguel A Brieño-Enríquez, Sergiy Libert
ABSTRACT
Food consumption is fundamental for life, and eating disorders often result in devastating or life-threatening conditions. Anorexia nervosa (AN) is characterized by a persistent restriction of energy intake, leading to lowered body weight, constant fear of gaining weight, and psychological disturbances of body perception. Herein, we demonstrate that SIRT1 inhibition, both genetically and pharmacologically, delays the onset and progression of AN behaviors in activity-based anorexia (ABA) models, while SIRT1 activation accelerates ABA phenotypes. Mechanistically, we suggest that SIRT1 promotes progression of ABA, in part through its interaction with NRF1, leading to suppression of a NMDA receptor subunit Grin2A. Our results suggest that AN may arise from pathological positive feedback loops: voluntary food restriction activates SIRT1, promoting anxiety, hyperactivity, and addiction to starvation, exacerbating the dieting and exercising, thus further activating SIRT1. We propose SIRT1 inhibition can break this cycle and provide a potential therapy for individuals suffering from AN.
Sigma-Aldrich
Anti-SIRT1 Antibody, clone 10E4, clone 10-E-4, from mouse