Merck
  • Chronic alcohol consumption alters extracellular space geometry and transmitter diffusion in the brain.
Chronic alcohol consumption alters extracellular space geometry and transmitter diffusion in the brain.
Science advances (2020-07-09)
Silvia De Santis, Alejandro Cosa-Linan, Raquel Garcia-Hernandez, Lesia Dmytrenko, Lydia Vargova, Ivan Vorisek, Serena Stopponi, Patrick Bach, Peter Kirsch, Falk Kiefer, Roberto Ciccocioppo, Eva Sykova, David Moratal, Wolfgang H Sommer, Santiago Canals
ABSTRACT
Already moderate alcohol consumption has detrimental long-term effects on brain function. However, how alcohol produces its potent addictive effects despite being a weak reinforcer is a poorly understood conundrum that likely hampers the development of successful interventions to limit heavy drinking. In this translational study, we demonstrate widespread increased mean diffusivity in the brain gray matter of chronically drinking humans and rats. These alterations appear soon after drinking initiation in rats, persist into early abstinence in both species, and are associated with a robust decrease in extracellular space tortuosity explained by a microglial reaction. Mathematical modeling of the diffusivity changes unveils an increased spatial reach of extrasynaptically released transmitters like dopamine that may contribute to alcohol's progressively enhanced addictive potency.
Sigma-Aldrich
Goat Anti-Mouse IgM Antibody, µ chain, Cy3 conjugate, Chemicon®, from goat
Sigma-Aldrich
Anti-Iba1/AIF1 Antibody, clone 20A12.1, from mouse