Merck
Light microscopy of proteins in their ultrastructural context.
Nature communications (2020-08-02)
Ons M'Saad, Joerg Bewersdorf
ABSTRACT
Resolving the distribution of specific proteins at the nanoscale in the ultrastructural context of the cell is a major challenge in fluorescence microscopy. We report the discovery of a new principle for an optical contrast equivalent to electron microscopy (EM) which reveals the ultrastructural context of the cells with a conventional confocal microscope. By decrowding the intracellular space through 13 to 21-fold physical expansion while simultaneously retaining the proteins, bulk (pan) labeling of the proteome resolves local protein densities and reveals the cellular nanoarchitecture by standard light microscopy.
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
TWEEN® 20, BioXtra, viscous liquid
Sigma-Aldrich
Sodium acetate buffer solution, pH 5.2±0.1 (25 °C), Molecular Biology, 3 M, 0.2 μm filtered
Sigma-Aldrich
Fibronectin bovine plasma, solution, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone B-5-1-2, ascites fluid
Sigma-Aldrich
N,N′-(1,2-Dihydroxyethylene)bisacrylamide, 97%
Sigma-Aldrich
Sodium borohydride, powder, ≥98.0%
Sigma-Aldrich
Atto 532 NHS ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Supelco
Tris(2-carboxyethyl)phosphine hydrochloride solution, 0.5 M, pH 7.0(aqueous solution; pH was adjusted with ammonium hydroxide)
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Triton X-100, Molecular Biology
Sigma-Aldrich
Sodium periodate, ACS reagent, ≥99.8%