Merck
  • NADPH oxidase 5α promotes the formation of CD271 tumor-initiating cells in oral cancer.
NADPH oxidase 5α promotes the formation of CD271 tumor-initiating cells in oral cancer.
American journal of cancer research (2020-07-10)
Wei Gao, Shaowei Xu, Minjuan Zhang, Shuai Liu, Sharie Pui-Kei Siu, Hanwei Peng, Judy Chun-Wai Ng, George Sai-Wah Tsao, Anthony Wing-Hung Chan, Velda Ling-Yu Chow, Jimmy Yu-Wai Chan, Thian-Sze Wong
ABSTRACT
Oral tongue squamous cell carcinoma (OTSCC) has a distinctive cell sub-population known as tumor-initiating cells (TICs). CD271 is a functional TIC receptor in head and neck cancers. The molecular mechanisms governing CD271 up-regulation remains unclear. Oxidative stress is a contributing factor in TIC development. Here, we explored the potential role of NADPH oxidase 5 (NOX5) and its regulatory mechanism on the development of CD271-expressing OTSCC. Our results showed that the splice variant NOX5α is the most prevalent form expressed in head and neck cancers. NOX5α enhanced OTSCC proliferation, migration, and invasion. Overexpression of NOX5α increased the size of OTSCC xenograft significantly in vivo. The tumor-promoting functions of NOX5α were mediated through the reactive oxygen species (ROS)-generating property. NOX5α activated ERK singling and increased CD271 expression at the transcription level. Also, NOX5α reduces the sensitivity of OTSCC to cisplatin and natural killer cells. The findings indicate that NOX5α plays an important part in the development of TIC in OTSCC.