Merck
  • Nanostructure generation during milk digestion in presence of a cell culture model simulating the small intestine.
Nanostructure generation during milk digestion in presence of a cell culture model simulating the small intestine.
Journal of colloid and interface science (2020-04-29)
Claudia Hempt, Mark Gontsarik, Tina Buerki-Thurnherr, Cordula Hirsch, Stefan Salentinig
ABSTRACT
The development of advanced oral delivery systems for bioactive compounds requires the fundamental understanding of the digestion process within the gastrointestinal tract. Towards this goal, dynamic invitro digestion models, capable of characterising the molecular as well as colloidal aspects of food, together with their biological interactions with relevant invitro cell culture models, are essential. In this study, we demonstrate a novel digestion model that combines flow-through time resolved small angle X-ray scattering (SAXS) with an invitro Caco-2/HT-29 cell co-culture model that also contained a mucus layer. This set-up allows the dynamic insitu characterisation of colloidal structures and their transport across a viable intestinal cell layer during simulated digestion. An integrated online SAXS - invitro cell co-culture model was developed and applied to study the digestion of nature's own emulsion, milk. The impact of the invitro cell culture on the digestion-triggered formation and evolution of highly ordered nanostructures in milk is demonstrated. Reported is also the crucial role of the mucus layer on top of the cell layer, protecting the cells from degradation by digestive juice components such as lipase. The novel model can open unique possibilities for the dynamic investigation of colloidal structure formation during lipid digestion and their effect on the uptake of bioactive molecules by the cells.
Sigma-Aldrich
meso-Tetraphenylporphyrin, BioReagent, suitable for fluorescence, ≥99.0% (HPLC)
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose, L-glutamine, and sodium bicarbonate, without sodium pyruvate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Penicillin − Streptomycin − Neomycin Solution Stabilized, formulated to contain ~5,000 units penicillin, 5 mg streptomycin and 10 mg neomycin/mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Trypsin-EDTA solution, 1 ×, sterile; sterile-filtered, BioReagent, suitable for cell culture, 0.5 g porcine trypsin and 0.2 g EDTA, 4Na per liter of Hanks′ Balanced Salt Solution with phenol red
Sigma-Aldrich
L-Glutamine solution, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, Heat Inactivated, non-USA origin, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Minimum Essential Medium Eagle, With Earle′s salts and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture